Skip to Content
Merck
All Photos(3)

Documents

06670

Sigma-Aldrich

2-Bromoethylamine hydrobromide

purum, ≥97.0% (AT)

Synonym(s):

2-Aminoethyl bromide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2NH2 · HBr
CAS Number:
Molecular Weight:
204.89
Beilstein:
3607605
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (AT)

form

crystalline

bp

181.3 °C/979.2 hPa

mp

170-175 °C (lit.)
170-175 °C

solubility

water: soluble 84.02 g/L at 28 °C
soluble

density

0.98 g/cm3 at 28.3 °C

SMILES string

Br.NCCBr

InChI

1S/C2H6BrN.BrH/c3-1-2-4;/h1-2,4H2;1H

InChI key

WJAXXWSZNSFVNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromoethylamine hydrobromide (BEA-HBr) can be used as a reactant in the preparation of:
  • Amino-functionalized ionic liquid, 1-aminoethyl-3-methylimidazolium hexafluorophosphate ([2-aemim][PF6]). [2-aemim][PF6] is employed as a catalyst in the synthesis of 4H-pyrans derivatives by treating with aromatic aldehydes, malononitrile, ethyl acetoacetate via Knoevenagel condensation reaction.
  • 2-(N-aryl-N-aroyl)amino-4,5-dihydrothiazole derivatives via cyclocondensation reaction.

It can be also employed as an alkylating agent for the surface modification of nylon to obtain primary/secondary/tertiary amine groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

142.9 °F - Pensky-Martens closed cup

Flash Point(C)

61.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino-functionalized ionic liquid as an efficient and recyclable catalyst for Knoevenagel reactions in water
Cai Y, et al.
Catalysis Letters, 109(1-2), 61-64 (2006)
Synthesis and QSAR Studies in 2-(N-aryl-N-aroyl) amino-4, 5-dihydrothiazole Derivatives as Potential Antithrombotic Agents
Saxena AK, et al.
Bioorganic & Medicinal Chemistry, 9(8), 2025-2034 (2001)
Nylon surface modification. Part 1. Targeting the amide groups for selective introduction of reactive functionalities
Jia X, et al.
Polymer, 47(14), 4916-4924 (2006)
G Bergström et al.
Journal of hypertension, 19(3 Pt 2), 659-665 (2001-05-01)
Restoring renal perfusion pressure (unclipping) of one-kidney-one-clip renal hypertensive (1 K1C) rats normalizes mean arterial pressure (MAP) rapidly. This has been attributed to salt/volume losses or release of the putative renal medullary depressor hormone (RMDH). To investigate the effects of
Samer Sourial et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 15(5), 424-435 (2010-05-25)
Currently there are no biomarkers for detecting collecting duct damage in man. Antibodies to several collecting duct-specific antigens exist but sandwich assays have been difficult to establish due to the need for two different antibodies to the same protein. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service