Skip to Content
Merck
All Photos(3)

Documents

142549

Sigma-Aldrich

3-Chloropropylamine hydrochloride

98%

Synonym(s):

1-Amino-3-chloropropane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
130.02
Beilstein:
3667451
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

145-150 °C (lit.)

SMILES string

Cl[H].NCCCCl

InChI

1S/C3H8ClN.ClH/c4-2-1-3-5;/h1-3,5H2;1H

InChI key

IHPRVZKJZGXTBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chloropropylamine hydrochloride was used in chemical treatment of carbon-enriched fly ash concentrates. It was used in the synthesis of linear, star and comb-like polyacrylamides by atomic transfer radical polymerization. It was used in the synthesis of halogen-functionalized aliphatic polyketones (macroinitiators).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer Molecular Architecture As a Tool for Controlling the Rheological Properties of Aqueous Polyacrylamide Solutions for Enhanced Oil Recovery.
Wever DAZ, et al.
Industrial & Engineering Chemistry Research, 52(47), 16993-17005 (2013)
M Mercedes Maroto-Valer et al.
Waste management (New York, N.Y.), 28(11), 2320-2328 (2007-12-21)
Fly ashes with high-unburned-carbon content, referred to as fly ash carbons, are an increasing problem for the utility industry, since they cannot be marketed as a cement extender and, therefore, have to be disposed. Previous work has explored the potential
Branched polyacrylamides: Synthesis and effect of molecular architecture on solution rheology.
Wever DAZ, et al.
Eur. Polymer J., 49(10), 3289-3301 (2013)
Huikang Yang et al.
International journal of biological macromolecules, 148, 483-492 (2020-01-12)
Redox-responsive theranostic nanoparticles based on poly-(N-ε-carbobenzyloxy-l-lysine) (PZLL) grafted hyaluronan (HA) (HA-g-SS-PZLL) copolymers were constructed for hepatocellular carcinoma diagnosis and therapy. These hyaluronan derivatives formed nanoparticles via a self-assembly process in aqueous solution at low concentration. Theranostic nanoparticles were obtained after
Laura Marcela Forero Ramirez et al.
Carbohydrate polymers, 245, 116457-116457 (2020-07-29)
Early detection of thrombotic events remains a big medical challenge. Dextran-based submicronic particles bearing Gd(DOTA) groups and functionalized with fucoidan have been produced via a simple and green water-in-oil emulsification/co-crosslinking process. Their capacity to bind to human activated platelets was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service