Skip to Content
Merck
All Photos(1)

Key Documents

30-4000

Sigma-Aldrich

Trichloroethylene

JIS special grade, ≥99.5%

Synonym(s):

TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
JIS special grade
Assay:
≥99.5%
bp:
86.7 °C (lit.)
vapor pressure:
61 mmHg ( 20 °C)

grade

JIS special grade

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

770 °F

availability

available only in Japan

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda L Eiden et al.
Journal of medical entomology, 52(3), 429-436 (2015-09-04)
Permethrin is a commonly used acaricide for tick control on domestic animals and in residential environments, while fipronil use is restricted to on-animal treatment. Following widespread reports of permethrin and fipronil application failures to control indoor infestations of Rhipicephalus sanguineus
Jo Philips et al.
Chemosphere, 91(1), 7-13 (2012-12-12)
Electron donor limitations likely reduce microbial enhanced trichloroethene (TCE) dense non-aqueous phase liquid (DNAPL) dissolution. This study quantitatively examined the relation between the DNAPL dissolution enhancement and the electron donor supply rate. An experiment used diffusion-cells with a 5.5 cm
Jun Yan et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1616), 20120320-20120320 (2013-03-13)
Dehalococcoides mccartyi strains are corrinoid-auxotrophic Bacteria and axenic cultures that require vitamin B12 (CN-Cbl) to conserve energy via organohalide respiration. Cultures of D. mccartyi strains BAV1, GT and FL2 grown with limiting amounts of 1 µg l(-1) CN-Cbl quickly depleted
Glinda S Cooper et al.
Environmental health perspectives, 117(5), 696-702 (2009-05-30)
Our objective was to examine experimental and epidemiologic studies pertaining to immune-related, and specifically autoimmune-related, effects of trichloroethylene (TCE). We performed a literature search of PubMed and reviewed bibliographies in identified articles. We then systematically reviewed immune-related data, focusing on
Cheryl Siegel Scott et al.
Environmental health perspectives, 114(9), 1471-1478 (2006-09-13)
A large body of epidemiologic evidence exists for exploring causal associations between cancer and trichloroethylene (TCE) exposure. The U.S. Environmental Protection Agency 2001 draft TCE health risk assessment concluded that epidemiologic studies, on the whole, support associations between TCE exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service