Skip to Content
Merck
All Photos(3)

Key Documents

251402

Sigma-Aldrich

Trichloroethylene

ACS reagent, ≥99.5%

Synonym(s):

TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04
grade:
ACS reagent
bp:
86.7 °C (lit.)
vapor pressure:
61 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

770 °F

dilution

(for analytical testing)

impurities

Free halogens, passes test
≤0.0001 meq/g Titr. acid
≤0.0003 meq/g Titr. base
≤0.02% water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

cation traces

heavy metals (as Pb): ≤1 ppm

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroethylene (TCE) is a halocarbon commonly used as an industrial chlorinated solvent. It can also be used as an intermediate in the production of other chemicals and refrigerants.

Application

Trichloroethylene can be used:
  • As a solvent in the C- and O-glycosylation reactions of tetrahydropyran acetals and 2-deoxyglucopyranosides to prepare SN2 products.
  • In the chemical industry for the synthesis of polyvinyl chloride, chloroacetic acid, hydrofluorocarbons, pharmaceuticals, insecticides, fungicides, fire retardants, fertilizer, and synthetic rubber.
  • As a starting material in the synthesis of dichloroacetylene.
  • As a reagent in the preparation of alkynic ethers.
  • As a reagent for ethynylation and vinylation process.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reductive dechlorination of trichloroethylene (TCE) in competition with Fe and Mn oxides-observed dynamics in H2-dependent terminal electron accepting processes.
Paul L, et al.
Geomicrobiology Journal (2015)
Eagleson M.
Concise Encyclopedia Chemistry, 332-332 (1994)
T M Vogel et al.
Applied and environmental microbiology, 49(5), 1080-1083 (1985-05-01)
Tetrachloroethylene (PCE) and trichloroethylene (TCE), common industrial solvents, are among the most frequent contaminants found in groundwater supplies. Due to the potential toxicity and carcinogenicity of chlorinated ethylenes, knowledge about their transformation potential is important in evaluating their environmental fate.
Eagleson M.
Concise Encyclopedia Chemistry, 215-215 (1994)
Jo Philips et al.
Chemosphere, 91(1), 7-13 (2012-12-12)
Electron donor limitations likely reduce microbial enhanced trichloroethene (TCE) dense non-aqueous phase liquid (DNAPL) dissolution. This study quantitatively examined the relation between the DNAPL dissolution enhancement and the electron donor supply rate. An experiment used diffusion-cells with a 5.5 cm

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. This product can be used for OIW analysis?

    1 answer
    1. This product is an ACS reagent grade trichloroethylene, typically used for synthetic purposes. This item is not tested or recommended for any specific analytical applications, such as oil-in-water analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service