Skip to Content
Merck
All Photos(1)

Key Documents

05-5230

Sigma-Aldrich

Cobalt(II) nitrate hexahydrate

JIS special grade, ≥98.0%

Synonym(s):

Cobaltous nitrate, Cobaltous nitrate hexahydrate, Nitric acid, cobalt(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
291.03
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: cobalt

availability

available only in Japan

pH

4 (20 °C, 100 g/L)

mp

55 °C (lit.)

SMILES string

O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Co.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

QGUAJWGNOXCYJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Llobet et al.
Revista espanola de fisiologia, 39(3), 291-298 (1983-09-01)
The acute toxicities of chloride, acetate, cobalt nitrate and cobalt sulphate (II) were investigated in rats. The values obtained for the LD50 (7 days) were 133, 194, 198 and 279 mg of Co/kg respectively, when the salts were given orally.
H H Chen et al.
Nucleic acids research, 10(11), 3561-3571 (1982-06-11)
Transient electric dichroism measurements have been used to observe the rotational relaxation times of 145 base pair fragments of poly (dGm5dC) and random sequence DNA to solution. From these the lengths of the fragments are calculated and the interbase pair
A time-resolved in situ quick-XAS investigation of thermal activation of Fischer-Tropsch silica-supported cobalt catalysts.
Jingping Hong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(10), 2802-2805 (2012-02-07)
William Buchmann et al.
Journal of mass spectrometry : JMS, 42(4), 517-526 (2007-02-27)
The electronebulization of a cobalt(II)/cysteine(Cys) mixture in water/methanol (50/50) produced mainly cobalt-cationized species. Three main groups of the Co-cationized species can be distinguished in the ESI-MS spectrum: (1) the cobalt complexes including the cysteine amino acid only (they can be
Massimiliano Aschi et al.
Journal of mass spectrometry : JMS, 44(7), 1038-1046 (2009-04-01)
The nature of the ionic species, formed by electrospray ionization (ESI) of Co(NO3)2/CH3OH solutions with a pair of aminoalcohols W and Y, has been investigated by mass spectrometric and computational methods. Collision induced dissociation (CID) of ions, formally corresponding to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service