Skip to Content
Merck
All Photos(1)

Key Documents

380024

Sigma-Aldrich

Iron(II) chloride tetrahydrate

99.99% trace metals basis

Synonym(s):

Ferrous chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2 · 4H2O
CAS Number:
Molecular Weight:
198.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
crystals and lumps

vapor pressure

10 mmHg ( 693 °C)

Quality Level

Assay

99.99% trace metals basis

form

crystals and lumps

impurities

≤150.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

SMILES string

O.O.O.O.Cl[Fe]Cl

InChI

1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

WSSMOXHYUFMBLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Iron(II) chloride tetrahydrate is a hydrated metallic halide. Crystal structure studies suggest that it crystallizes in monoclinic system having space group P21/c.

Application

A variety of organically templated iron phosphates, of interest because of their rich crystal chemistry and their ability to absorb large molecules in their micropores,were prepared using this compound.
Iron(II) chloride tetrahydrate (Ferrous chloride tetrahydrate, FeCl2·4H2O) has been used in the preparation of bentonite/iron nanoparticles.
It may be used in the following studies:
  • Preparation of composites of multiwall carbon nanotubes (MWCNTs)/nano-iron oxide.
  • Preparation of magnetite (Fe3O4) nanoparticles.
  • Solid-phase synthesis of [Fe(phen)3]Cl2·nH2O (phen = 1,10-phenanthroline).4

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bibek Thapa et al.
Scientific reports, 9(1), 5633-5633 (2019-04-06)
The engineering of materials with controlled magnetic properties by means other than a magnetic field is of great interest in nanotechnology. In this study, we report engineered magnetic graphene oxide (MGO) in the nanocomposite form of iron oxide nanoparticles (IO)-graphene
Synthesis and characterization of bentonite/iron nanoparticles and their application as adsorbent of cobalt ions.
Shahwan T, et al.
Applied Clay Science, 47(3), 257-262 (2010)
Preparation and characterization of magnetic chitosan nanospheres.
Hritcu D, et al.
Turkish Journal of Chemistry, 33, 785-796 (2009)
The crystal structure of iron (II) chloride tetrahydrate.
Penfold BR and Grigor JA.
Acta Crystallographica, 12(11), 850854-850854 (1959)
Chromium removal by combining the magnetic properties of iron oxide with adsorption properties of carbon nanotubes.
Gupta VK, et al.
Water Research, 45(6), 2207-2212 (2011)

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service