Skip to Content
Merck
All Photos(1)

Key Documents

11089161001

Roche

Streptavidin-AP conjugate

solution, pkg of 1 mL (1,000U)

Synonym(s):

streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

solution

Quality Level

packaging

pkg of 1 mL (1,000U)

manufacturer/tradename

Roche

storage temp.

2-8°C

Related Categories

General description

Streptavidin is covalently coupled to alkaline phosphatase (AP) for the preparation of streptavidin-AP-conjugate.

Alkaline Phosphatase
Alkaline Phosphatase is often used as a label in enzyme immunoassays. AP converts the sensitive substrate X-phosphate/NBT into a stable, water soluble product.

Application

Streptavidin-AP has been used to detect biotinylated compounds using:
  • ELISA[1]
  • Immunohistocytochemistry
  • Western blot[2][3]

Physical form

Solution, stablilized in 50 mM Triethanolamine buffer, 3 M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2, 10 mg/ml bovine serum albumin, pH 7.6.

Preparation Note

Working solution: 50 - 1,000 mU/ml (i.e., stock solution can be diluted). Dilute the conjugate stock solution in 100 mM Tris-HCl, 150 mM NaCl, pH 7.5.

Working concentration: Working concentration of conjugate will depend on the application and substrate. Indicated concentrations should be taken as a guideline.

ELISA: 50 - 200 mU/ml (1:5000 - 1:20,000 dilution)
Immunohisto/cytochemistry: 250 - 1,000 mU/ml (1:1000 - 1:4000 dilution)
Western blot: 200 - 500 mU/ml (1:2000 - 1:5000 dilution)

Stability: The concentrated stock solution is stable when stored at 2 to 8°C until the expiration date printed on the label. Do not freeze!

Other Notes

For life science research only. Not for use in diagnostic procedures.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ablation of BMP signaling hampers the blastema formation in Poecilia latipinna by dysregulating the extracellular matrix remodeling and cell cycle turnover
Patel S, et al.
Zoology (Jena, Germany), 133, 17-26 (2019)
Johan L K Van Hove et al.
Molecular genetics and metabolism, 95(4), 201-205 (2008-11-01)
We investigated in a patient with holocarboxylase synthetase deficiency, the relation between the biochemical and genetic factors of the mutant protein with the pharmacokinetic factors of successful biotin treatment. A girl exhibited abnormal skin at birth, and developed in the
Manuela Sauter et al.
STAR protocols, 3(3), 101664-101664 (2022-09-14)
Different types of immune cells are involved in atherogenesis and may act atheroprotective or atheroprogressive. Here, we describe an in vitro approach to analyze CD11c+ cells and CD11c+-derived ApoE in atherosclerosis. The major steps include harvesting mouse bone marrow, plating
M Degerman Gunnarsson et al.
Dementia and geriatric cognitive disorders, 29(3), 204-212 (2010-03-25)
The positron emission tomography (PET) radiotracer Pittsburgh Compound-B (PIB) is an in vivo ligand for measuring beta-amyloid (Abeta) load. Associations between PET PIB and cerebrospinal fluid (CSF) Abeta1-42 and apolipoprotein E epsilon4 (APOE epsilon4) have been observed in several studies
Manuela Sauter et al.
iScience, 25(1), 103677-103677 (2022-01-18)
Atherosclerosis is studied in models with dysfunctional lipid homeostasis-predominantly the ApoE-/- mouse. The role of antigen-presenting cells (APCs) for lipid homeostasis is not clear. Using a LacZ reporter mouse, we showed that CD11c+ cells were enriched in aortae of ApoE-/-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service