Skip to Content
Merck
All Photos(1)

Documents

11089153001

Roche

Streptavidin-POD conjugate

liquid, pkg of 1 mL

Synonym(s):

streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

Quality Level

packaging

pkg of 1 mL

manufacturer/tradename

Roche

storage temp.

-10 to -25°C

General description

Streptavidin is covalently coupled to horseradish peroxidase. The conjugate is used for detection of biotin-labelled substances.

Application

Streptavidin-POD has been used for:
  • ELISA
  • In situ hybridization
  • Western blot
It is also used for immunohistochemistry.

Unit Definition

One unit Streptavidin-POD, corresponding to one unit peroxidase, will oxidize 1 μmol of guaiacol tetramer in one minute at +25 °C and pH 7.0.

Unit Conversion: 1 U (+25 °C; guaiacol as substrate; pH7.0) corresponds to 1.24 U (+25 °C; 1.7 mM ABTS as substrate; pH= 6.0) or to 4.0 U (+25 °C; 8.7 mM ABTS as substrate; pH = 5.0).

Physical form

Liquid, stabilized

Preparation Note

Working concentration: 5 - 500 mU/ml (i.e., stock solution can be diluted).Working concentration of conjugate depends on application and substrate. The following concentrations should be taken as a guideline:
  • ELISA: 50 to 200 mU/ml
  • Immunohistocytochemistry: 250 to 1000 mU/ml
  • Western blot: 200 to 500 mU/ml

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelika Hammerer-Lercher et al.
Clinical chemistry, 54(5), 858-865 (2008-03-15)
The specific forms of pro-B-type natriuretic peptide (proBNP) that occur in human blood are not yet clear. We demonstrated the presence of several proBNP forms in human plasma with a new affinity chromatography method that can be used in combination
Benjamin Meyer et al.
Frontiers in immunology, 13, 841009-841009 (2022-05-20)
To comprehensively evaluate SARS-CoV-2 specific B-cell and antibody responses up to one year after mild COVID-19. In 31 mildly symptomatic COVID-19 participants SARS-CoV-2-specific plasmablasts and antigen-specific memory B cells were measured by ELISpot. Binding antibodies directed against the proteins spike
Triple-Labeling Whole-Mount In Situ Hybridization Method for Analysis of Overlapping Gene Expression in Brain Tissue with High Level of Autofluorescence.
Chen J
Journal of Cytology & Histology, http://dx-http://dx (2015)
BAP1 and PTEN mutations shape the immunological landscape of clear cell renal cell carcinoma and reveal the intertumoral heterogeneity of T cell suppression: a proof-of-concept study.
Friedhoff, et al.
Cancer Immunology, Immunotherapy, 72, 1603-1618 (2023)
Husheem Michael et al.
Frontiers in immunology, 11, 196-196 (2020-03-03)
Human rotavirus (HRV) is a leading cause of morbidity and mortality in children, especially in developing countries. Malnutrition is prevalent in these countries, which may contribute to the decreased oral vaccine efficacy, posing a concern for global health. Neonatal gnotobiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service