Skip to Content
Merck
All Photos(5)

Key Documents

G6378

Sigma-Aldrich

Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae)

Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)

Synonym(s):

G-6-P-DH, Zwischenferment

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type XV

Quality Level

form

lyophilized powder

specific activity

200-400 units/mg protein (modified Warburg-Christian)

mol wt

128 kDa

purified by

crystallization

application(s)

diagnostic assay manufacturing

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Glucose 6-phosphate dehydrogenase (G-6-P-DH) is a key regulatory enzyme in the first step of the pentose phosphate pathway. G-6-P-DH is a glycoprotein with a molecular mass of 128 kDa (gel filtration).

Application

Glucose-6-phosphate dehydrogenase is used to test ketose reductase activity in developing maize endosperm.

Biochem/physiol Actions

Glucose-6-phosphate dehydrogenase catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway<<<17>>>.
Glucose-6-phosphate dehydrogenase catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway.

Unit Definition

One unit will oxidize 1.0 μmole of D-glucose 6-phosphate to 6-phospho-D-gluconate per min in the presence of NADP at pH 7.4 at 25 °C.

Physical form

Lyophilized and essentially sulfate-free; contains approx. 20% sodium citrate based on dry weight

Preparation Note

Prepared from G7877

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D C Doehlert
Plant physiology, 84(3), 830-834 (1987-07-01)
Ketose reductase (NAD-dependent polyol dehydrogenase EC 1.1.1.14) activity, which catalyzes the NADH-dependent reduction of fructose to sorbitol (d-glucitol), was detected in developing maize (Zea mays L.) endosperm, purified 104-fold from this tissue, and partially characterized. Product analysis by high performance
K E Reilly et al.
Biochemical and biophysical research communications, 216(3), 993-998 (1995-11-22)
A commercial preparation of glucose-6-phosphate dehydrogenase (G6PD) purified from Saccharomyces cerevisiae was subjected to PAGE analysis under both nondenaturing and denaturing conditions. The enzyme, identified by both activity staining and anti-yeast G6PD antibody immunoblotting, was shown to contain carbohydrate using
Yong Yang et al.
Frontiers in microbiology, 9, 1428-1428 (2018-07-20)
Mycobacteria spontaneously form surface-associated multicellular communities, called biofilms, which display resistance to a wide range of exogenous stresses. A causal relationship between biofilm formation and emergence of stress resistance is not known. Here, we report that activation of a nitrogen
P Andrews
The Biochemical journal, 96(3), 595-606 (1965-09-01)
1. Correlation between elution volume, V(e), and molecular weight was investigated for gel filtration of proteins of molecular weights ranging from 3500 (glucagon) to 820000 (alpha-crystallin) on Sephadex G-200 columns at pH7.5. 2. Allowing for uncertainties in the molecular weights
Sean Rogers et al.
eLife, 10 (2021-04-08)
Eukaryotes compartmentalize metabolic pathways into sub-cellular domains, but the role of inter-organelle contacts in organizing metabolic reactions remains poorly understood. Here, we show that in response to acute glucose restriction (AGR) yeast undergo metabolic remodeling of their mevalonate pathway that

Protocols

To measure glucose-6-phosphate dehydrogenase activity, beta-nicotinamide adenine dinucleotide phosphate is used in a spectrophotometric rate determination assay at 340 nm.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service