Skip to Content
Merck
All Photos(2)

Key Documents

MABE1045

Sigma-Aldrich

Anti-CTIP2/BCL11B Antibody, clone 25B6

Anti-CTIP2/BCL11B Antibody, clone 25B6
1 of 1 reviewers received a sample product or took part in a promotion

clone 25B6, from rat

Synonym(s):

B-cell lymphoma/leukemia 11B, B-cell CLL/lymphoma 11B, BCL-11B, COUP-TF-interacting protein 2, mRit1, Radiation-induced tumor suppressor gene 1 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

25B6, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... BCL11B(64919)

General description

B-cell lymphoma/leukemia 11B (UniProt Q99PV8; also known as B-cell CLL/lymphoma 11B, BCL-11B, COUP-TF-interacting protein 2, mRit1, Radiation-induced tumor suppressor gene 1 protein) is encoded by the Bcl11b (also known as Ctip2, Rit1) gene (Gene ID 58208) in murine species. CTIP2/CBL11B is a C2H2 zinc finger protein that binds GC-rich response elements and is reported to associate with a variety of cofactors, including the corepressor complexes NuRD, Sirt1, histone acetyltransferase (HAT) p300, functioning both as a transcriptional repressor and activator. CTIP2/CBL11B is required at multiple developmental checkpoints for T cell commitment and maintenance. CTIP2/CBL11B expression is initiated at double-negative 2 (DN2) stage of T cell development and is maintained in mature T lymphocytes, with lower levels found in NK cells, but not in B cells, myeloid or dendritic cells. Bcl11b-/- mice die shortly after birth due to a blockade of thymocyte development at the DN2 stage. Bcl11b-deficient DN2 thymocytes are highly proliferative and retain the potential to differentiate into NK cells and myeloid lineages, but not mature T-cells.

Specificity

Clone 25B6 recognizes both the Alpha and Beta spliced isoforms of human and mouse CTIP2/BCL11B.

Immunogen

GST-tagged recombinant full-length mouse CTIP2 Beta isoform (Senawong, T., et al. (2003). J. Biol. Chem. 278(44):43041-43050).

Application

Anti-CTIP2/BCL11B Antibody, clone 25B6, Cat. No. MABE1045, is a highly specific rat monoclonal antibody that targets CTIP2 and has been tested in Immunofluorescence, Immunohistochemistry, and Western Blotting.
Immunohistochemistry Analysis: A 1:240 dilution from a representative lot detected CTIP2/BCL11B in mouse brain tissue sections.

Immunofluorescence Analysis: A representative lot detected CTIP2 localization in 4% paraformaldehyde-fixed floating embryonic (E14 & E16) and postnatal (P3 & P6) mouse brain sections (Arlotta, T., et al. (2005). Neuron. 45(2):207-221).

Western Blotting Analysis: A representative lot detected doublet CTIP2 bands, representing two spliced isoforms, in Jurkat nuclear extracts as well as in SIRT1 immunoprecipitate (Senawong, T., et al. (2003). J. Biol. Chem. 278(44):43041-43050).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CTIP2/BCL11B in 10 µg of HEK293 cell lysate.

Target description

~96 kDa observed. 95.52/94.57 kDa (human/mouse isoform Alpha) and 88.48/87.34 kDa (human/mouse isoform Beta) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified rat monoclonal antibody IgG2a in buffer containing 150 mM NaCl, 15 mM HEPES, pH 7.5, with 0.02% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Munz et al.
Cell, 186(9), 1930-1949 (2023-04-19)
Cortical circuits are composed predominantly of pyramidal-to-pyramidal neuron connections, yet their assembly during embryonic development is not well understood. We show that mouse embryonic Rbp4-Cre cortical neurons, transcriptomically closest to layer 5 pyramidal neurons, display two phases of circuit assembly
Philippe Foerster et al.
Development (Cambridge, England), 144(2), 201-210 (2016-12-21)
Radial glial cells (RCGs) are self-renewing progenitor cells that give rise to neurons and glia during embryonic development. Throughout neurogenesis, these cells contact the cerebral ventricles and bear a primary cilium. Although the role of the primary cilium in embryonic
Gokhan Dalgin et al.
Scientific reports, 11(1), 21590-21590 (2021-11-05)
The gene KCNJ11 encodes Kir6.2 a major subunit of the ATP-sensitive potassium channel (KATP) expressed in both the pancreas and brain. Heterozygous gain of function mutations in KCNJ11 can cause neonatal diabetes mellitus (NDM). In addition, many patients exhibit neurological

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

    • Review 1
    • Vote 1
    5 out of 5 stars.

    Anti-CTIP2/BCL11B Antibody

    Used for IF on mouse brain sections. Very clean and strong signal.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service