Skip to Content
Merck
All Photos(6)

Key Documents

MAB397

Sigma-Aldrich

Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4

clone 6C4, Chemicon®, from mouse

Synonym(s):

GluR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C4, monoclonal

species reactivity

monkey

species reactivity (predicted by homology)

mouse, rat, canine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

dog ... Gria2(482667)
mouse ... Gria2(14800)
rat ... Gria2(29627)
rhesus monkey ... Gria2(574344)

General description

Glutamate receptors (GluRs) can be categorized as ionotropic or metabotropic and subcatergorized by their agonist preferences (NMDA, AMPA or Kainic acid). There are four types of AMPA selective GluR subunits (GluR1, GluR2, GluR3 and GluR4). Tetrameric or pentameric combinations of different subunits contributes to the functional diversity of AMPA receptors. In general, AMPA receptors mediate fast synaptic current at most excitatory synapses, with stoichiometry characterized by subtype composition. Although subunit composition of AMPA receptors varies, they must contain at least one edited GluR2 subunit to be calcium impermeable. The critical residue controlling calcium permeability is in the pore loop region. In GluR1, GluR3, and GluR4, this positionis occupied by a Gln residue. In GluR2, it is occupied by an Arg residue. It has been shown experimentally that the presence of Arg in this position blocks CA2+ ion permeability, while a Gln does not. Relative calcium permeability in AMPA receptor channels may be significant in pathological neurotoxic damage and long term changes in nervous system responses.

Specificity

Recognizes the large N-terminal extracellular domain of Glutamate Receptor 2 (GluR2). No cross-reactivity observed with other AMPA/Kainate GluR subunits. On western blots of brain extracts from rat, macaque monkey, and dog, MAB397 recognizes a band at approximately 102 kDa corresponding to full length GluR2. Other proteins noted by Western blot at 66 kDa or lower molecular weight appear to be breakdown products of GluR2 (Vissavajjhala, 1996). By immunohistochemistry GluR2 is widely distributed at both the cellular and synaptic levels. MAB397 recognizes GluR2 present in a vast majority of, but not all, GABAergic interneurons (Vissavajjhala, 1996).

Application

Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4 detects level of Glutamate Receptor 2 & has been published & validated for use in ELISA, IC, IH, IP, RIA & WB with more than 50 product citations.
Immunocytochemistry:
on 4% paraformaldehyde fixed cells was used in a previous lot:2-3 µg/mL (Vissavajjhala, 1996; Osten, 1998; Passafaro, 2001).

ELISA/RIA:
A previous lot of this antibody was used in ELISA/RIA.(Vissavajjhala, 1996).

Immunohistochemistry on 50 µm, 4% paraformaldehyde fixed brain sections: 1:500-1:800 (Vissavajjhala, 1996; Yung, 1998; Kumar; 2002).

Immunoprecipitation: 2-4 µg/mL (Osten, 1998).

Western blot: 1-2 µg/mL (Vissavajjhala, 1996; Osten, 1998); membrane preparations are suggested for enhanced signals.

Optimal working dilutions and protocols must be determined by end user.

Quality

Routinely evaluated by Western Blot on mouse brain lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected glutamate receptor 2 on 10 μg of mouse brain lysates.

Target description

102 kDa

Physical form

Format: Purified
Purified immunoglobulin from culture supernatant
Purified mouse immunoglobin IgG2a liquid in buffer containing PBS, no preservative.

Storage and Stability

Stable for 6 months at -20ºC in undiluted aliquots from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage the IgG2a and affect product performance.

Analysis Note

Control
Positive Control: Cerebral cortex and pyramidal neurons, mouse brain lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyrosine phosphatases regulate AMPA receptor trafficking during metabotropic glutamate receptor-mediated long-term depression.
Moult, PR; Gladding, CM; Sanderson, TM; Fitzjohn, SM; Bashir, ZI; Molnar, E; Collingridge, GL
The Journal of Neuroscience null
S-SCAM/MAGI-2 is an essential synaptic scaffolding molecule for the GluA2-containing maintenance pool of AMPA receptors.
Danielson, E; Zhang, N; Metallo, J; Kaleka, K; Shin, SM; Gerges, N; Lee, SH
The Journal of Neuroscience null
Melanie A Gainey et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), E3590-E3599 (2015-06-26)
Synaptic scaling is a form of homeostatic plasticity that stabilizes neuronal firing in response to changes in synapse number and strength. Scaling up in response to action-potential blockade is accomplished through increased synaptic accumulation of GluA2-containing AMPA receptors (AMPAR), but
Translamellar disinhibition in the rat hippocampal dentate gyrus after seizure-induced degeneration of vulnerable hilar neurons.
Zappone, CA; Sloviter, RS
The Journal of Neuroscience null
Differential regulation of AMPA receptor and GABA receptor trafficking by tumor necrosis factor-alpha.
Stellwagen, D; Beattie, EC; Seo, JY; Malenka, RC
The Journal of Neuroscience null

Questions

  1. Is it possible to use two primary antibodies from different species but of the same isotype for double staining Immunofluorescence? Specifically, considering MAB397 and GluR2.

    1 answer
    1. Yes, it is possible to use two primary antibodies from different species but of the same isotype for double staining Immunofluorescence. Specifically, for MAB397 and GluR2, one can use an anti-mouse and an anti-rabbit secondary antibody to distinguish reactivity of the primary antibodies.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service