Skip to Content
Merck
All Photos(8)

Key Documents

AB144P

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

CHEMICON®, goat polyclonal

Synonym(s):

Choline acetylase, acetyl CoA:choline O-acetyltransferase, choline acetyltransferase, ChAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, opossum, guinea pig, rat, mouse, zebrafish (THE JOURNAL OF COMPARATIVE NEUROLOGY 474:75–107 (2004).), avian, monkey, chicken

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

chicken ... Chat(395314)
human ... CHAT(1103)
mouse ... Chat(12647)
rat ... Chat(290567)
rhesus monkey ... Chat(709977)

General description

Choline acetyltransferase (ChAT) has a molecular weight of about 68 kDa. It is synthesized in the parakaryon and is transported to the axon terminals. ChAT belongs to the choline/carnitine acyltransferase family. It seems that only cholinergic neurons have significant amounts of ChAT making anti-choline acetyltransferase a useful specific marker. Choline acetyltransferase is found in cholinergic neurons in brain and spinal cord.

Specificity

Expected to cross-react with Avian, chicken, guinea pig, monkey, rat, and opposum.

Immunogen

Human placental enzyme.

Application

Anti-Choline Acetyltransferase Antibody has been used in:
  • immunohistochemical staining
  • western blotting
  • immunocytochemistry

Immunohistochemistry:
A previous lot of this antibody was used at 1:100.

Optimal working dilutions and protocols must be determined by end user.

Biochem/physiol Actions

Choline acetyltransferase (CHAT) is involved in the synthesis of acetylcholine in neurons. It catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses. CHAT catalyzes the transfer of an acetyl group between acetyl-coenzyme A and choline. Mutations in the gene encoding this protein have been associated with congenital neuromuscular disorders.

Quality

Routinely evaluated by Western Blot on mouse brain lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected CHAT on 10 μg of mouse brain lysates.

Target description

70/74 kDa

Physical form

Affinity purified
Purified goat polyclonal in buffer containing 5 mg/mL BSA and 0.2% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 1 year from date of receipt. Avoid repeated freeze/thaw cycles.
For short term storage (3-4 weeks) maintain at 2-8°C.

Analysis Note

Control
Human placenta lysates, rat forebrain/rostral hypothalamus.

Other Notes

Research Category:
Neuroscience

Research Sub Category:
Neurotransmitters & Receptors

Neuronal & Glial Markers

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasticity of motor network and function in the absence of corticospinal projection
Han Q, et al.
Experimental Neurology, 194-208 (2015)
The human choline acetyltransferase gene encodes two proteins
Grosman DD, et al.
Journal of Neurochemistry (1995)
Blood pressure regulation by CD4+lymphocytes expressing choline acetyltransferase
Olofsson PS, et al.
Nature Biotechnology, 34(10), 1066-1071 (2016)
S D Motts et al.
Neuroscience, 160(1), 103-114 (2009-03-14)
We combined retrograde tracing with immunohistochemistry for choline acetyltransferase to identify the source of cholinergic input to the inferior colliculus (IC) in guinea pigs. Injection of a retrograde tracer into one IC labeled cells in many brainstem nuclei. Retrogradely-labeled cells
Choline acetyltransferase structure reveals distribution of mutations that cause motor disorders
Cai Y, et al.
The Embo Journal, 2047-2047 (2004)

Articles

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

See All

Questions

  1. Bonjour, quel est l'isotype de cet anticorps ?

    1 answer
    1. This antibody is an IgG isotype. The whole protein was used for immunization and the epitope has not been mapped. This is a polyclonal antibody and can bind to more than one region.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service