Skip to Content
Merck
All Photos(1)

Documents

MAB8744

Sigma-Aldrich

Anti-St. Louis Encephalitis Antibody, MSI-7, clone 6B6C-1

clone 6b6c-1, Chemicon®, from mouse

Synonym(s):

SLE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

6b6c-1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
inhibition assay: suitable (hemagglutination)

isotype

IgG2a

shipped in

wet ice

Specificity

Cross reactivity to St. Louis encephalitis (MSI-7), Japanese encephalitis (Nakayama), West Nile (EG 101), Murray Valley encephalitis (Original), Yellow fever (17D), Dengue 1 (Hawaii) Dengue 2 (New Guinea C), Dengue 3 (H87) and Dengue 4 (H241) is determined by ELISA.

Immunogen

Epitope: MSI-7

Application

Anti-St. Louis Encephalitis Antibody, MSI-7, clone 6B6C-1 detects level of St. Louis Encephalitis & has been published & validated for use in ELISA, HI & IF.
ELISA

Indirect immunofluorescence Hemagglutination inhibition

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Patient culture

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exclusion of West Nile virus superinfection through RNA replication.
Zou, G; Zhang, B; Lim, PY; Yuan, Z; Bernard, KA; Shi, PY
Journal of virology null
Comparative evaluation of permissiveness to dengue virus serotype 2 infection in primary rodent macrophages.
Prada-Arismendy, J; Rincon, V; Castellanos, JE
Journal of tropical medicine null
Isolation of saint louis encephalitis virus from a horse with neurological disease in Brazil.
Rosa, R; Costa, EA; Marques, RE; Oliveira, TS; Furtini, R; Bomfim, MR; Teixeira et al.
PLoS Neglected Tropical Diseases null
J T Roehrig et al.
Journal of clinical microbiology, 35(7), 1887-1890 (1997-07-01)
We used previously characterized murine monoclonal antibodies to develop a panel useful in subtyping Venezuelan equine encephalitis (VEE) viruses by an indirect fluorescent antibody assay. This panel worked well with either prototype VEE viruses or a series of more recent
Jaime E Castellanos et al.
Biomedica : revista del Instituto Nacional de Salud, 36(0), 156-158 (2016-09-14)
Dengue is a human disease caused by a virus with the same name, which is transmitted by the bite of Aedes mosquitoes. The infection has a wide range of clinical presentations ranging from asymptomatic to fatal cases, with the pediatric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service