Skip to Content
Merck
All Photos(1)

Key Documents

8.03131

Sigma-Aldrich

Diethylene glycol

Technipur®, for synthesis

Synonym(s):

2,2′-Oxydiethanol, Bis(2-hydroxyethyl) ether, 2-Hydroxyethyl ether, Diglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2O
CAS Number:
Molecular Weight:
106.12
Beilstein:
969209
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.05
grade:
for synthesis
bp:
245 °C (lit.)
vapor pressure:
0.01 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor density

2.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

autoignition temp.

442 °F

expl. lim.

2-12.3 %

refractive index

n20/D 1.447 (lit.)

pH

6 (20 °C, 200 g/L in H2O)

bp

245 °C (lit.)

mp

-6.5 °C (DIN )
−10 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

SMILES string

OCCOCCO

InChI

1S/C4H10O3/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

MTHSVFCYNBDYFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylene glycol is widely used as a solvent and ingredient for many consumer products like antifreeze preparations, wallpaper strippers, brake fluids, and artificial fog solutions.[1]

Application

Diethylene glycol can be used:
  • To improve the digital inkjet printability of dye solutions on cotton fabrics.[2]
  • As a humectant in the pigment ink formulation applicable in inkjet printing.[3]
  • As a solvent in the synthesis of Bi2S3 nanorod applicable as a direct band-gap semiconductor in thermoelectric devices, photoelectric devices.[4]

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of nano-coated pigment ink formulation on ink-jet printability and printing accuracy
Li M, et al.
Coloration Technology, 133(6), 476-484 (2017)
Marraffa JM, et al. Diethylene glycol: widely used solvent presents serious poisoning potential.
Marraffa JM, et al
The Journal of Emergency Medicine, 35(4), 401-406 (2008)
Synthesis and characterization of Bi2S3 nanorods by solvothermal method in polyol media
Zhang H and Wang L
Materials Letters, 61( 8-9), 1667-1670 (2007)
Effect of Diethylene Glycol on the Inkjet Printability of Reactive Dye Solution for Cotton Fabrics
Hui Peng H, et al.
Langmuir, 37(4), 1493-1500 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service