Skip to Content
Merck
All Photos(1)

Key Documents

P4308

Sigma-Aldrich

Potassium hexahydroxoantimonate(V)

Synonym(s):

Potassium antimonate, hydrated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSb(OH)6
CAS Number:
Molecular Weight:
262.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

concentration

44.3-49.8% Sb (iodometric)

SMILES string

[K+].O[Sb-](O)(O)(O)(O)O

InChI

1S/K.6H2O.Sb/h;6*1H2;/q+1;;;;;;;+5/p-6

InChI key

DUBGVNNAYGFUKU-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Potassium hexahydroxoantimonate(V) can be used as a precursor salt to synthesize:
  • Antimony oxide (Sb2O3), which is applicable as a sorbent material for the removal of cobalt from radioactive waste.
  • Amphiphilic antimony(V)complexes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flaviana R Fernandes et al.
Antimicrobial agents and chemotherapy, 57(9), 4229-4236 (2013-06-26)
The need for daily parenteral administration is an important limitation in the clinical use of pentavalent antimonial drugs against leishmaniasis. In this study, amphiphilic antimony(V) complexes were prepared from alkylmethylglucamides (L8 and L10, with carbon chain lengths of 8 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service