Skip to Content
Merck
All Photos(3)

Key Documents

I7780

Sigma-Aldrich

Iodoethane

contains copper as stabilizer, ReagentPlus®, 99%

Synonym(s):

Ethyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5I
CAS Number:
Molecular Weight:
155.97
Beilstein:
505934
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

5.38 (vs air)

Quality Level

vapor pressure

100 mmHg ( 18 °C)

product line

ReagentPlus®

Assay

99%

contains

copper as stabilizer

refractive index

n20/D 1.513 (lit.)

bp

69-73 °C (lit.)

mp

−108 °C (lit.)

density

1.940 g/mL at 25 °C (lit.)

SMILES string

CCI

InChI

1S/C2H5I/c1-2-3/h2H2,1H3

InChI key

HVTICUPFWKNHNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodoethane also known as ethyl iodide is an alkyl iodide. The interaction of iodoethane with Pt(111) surfaces has been analyzed by using thermal desorption and X-ray photoelectron spectroscopy.[1] Its microwave spectrum has been recorded and studied.[2] The dynamics of the gas phase photolysis of iodoethane have been studied using resonance Raman spectra.[3] Its thermodynamic characteristics in the ideal state have been reported.[4]

Application

Iodoethane has been used for performing gas-phase experiments for recording extreme ultraviolet (XUV) photoelectron spectra.[5]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup - (External MSDS)

Flash Point(C)

53 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Xu et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-15)
Preparation of aluminosilicate ferrierite (FER) zeolite nanosheets with controllable thickness in the presence of a sole organic ammonium is attractive, but still challenging. In this report, with the employment of N,N-diethyl-cis-2,6-dimethylpiperidinium (DMP) as both a structure directing agent and crystal
Formation and thermal decomposition of ethyl groups on transition metal surfaces: Ethyl iodide on Pt (111).
Zaera F.
Surface Science, 219(3), 453-466 (1989)
Substituent effects on gas-phase photodissociation dynamics: resonance Raman spectra of ethyl iodide, isopropyl iodide, and tert-butyl iodide.
Phillips DL, et al.
The Journal of Physical Chemistry, 95(23), 9085-9091 (1991)
Microwave Spectrum of Ethyl Iodide, I.
Kasuya T and Oka T.
Journal of the Physical Society of Japan, 15(2), 296-303 (1960)
Yogesh Gawale et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(27), 6570-6578 (2017-02-23)
Four new N-ethylcarbazole-linked aza-boron-dipyrromethene (aza-BODIPY) dyes (8 a,b and 9 a,b) were synthesized and characterized. The presence of the N-ethylcarbazole moiety shifts their absorption and fluorescence spectra to the near-infrared region, λ≈650-730 nm, of the electromagnetic spectrum. These dyes possess strong molar absorptivity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service