Skip to Content
Merck
All Photos(1)

Key Documents

8.00457

Sigma-Aldrich

Butyric acid

for synthesis

Synonym(s):

Butyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-532-3
NACRES:
NA.22

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

0.56 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

425 °C

potency

2940 mg/kg LD50, oral (Rat)
509 mg/kg LD50, skin (Rabbit)

expl. lim.

2-12.3 % (v/v)

pH

2.5 (20 °C, 100 g/L in H2O)

bp

162-165 °C/1013 mbar

mp

-8--5 °C

transition temp

flash point 69 °C

density

0.96 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCC

InChI

1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)

InChI key

FERIUCNNQQJTOY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.956 - 0.958
Water (K. F.): ≤ 0.2 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service