Skip to Content
Merck
All Photos(1)

Key Documents

12-448

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate

Upstate®, from goat

Synonym(s):

Anti-Rabbit IgG, Goat Anti-Rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
This Goat anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate is validated for use in ELISA, WB, IH for the detection of Rabbit IgG.

Quality

Routinely evaluated for use as a secondary antibody in WB, ELISA and IHC.

Physical form

50mM Tris chloride, pH 8.0, 150mM NaCl, 1mM MgCl, 0.1mM ZnCl, 0.01% (w/v) sodium azide, 50% (v/v) glycerol and 10mg/ml BSA

Storage and Stability

Stable for 1 year at 4°C from date of shipment. Liquid at 4°C. DO NOT FREEZE.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intron retention in the Drosophila melanogaster Rieske Iron Sulphur Protein gene generated a new protein.
Gontijo, AM; Miguela, V; Whiting, MF; Woodruff, RC; Dominguez, M
Nature Communications null
Angiotensin II receptor expression and relation to Helicobacter pylori-infection in the stomach of the Mongolian gerbil.
Hallersund, P; Helander, HF; Casselbrant, A; Edebo, A; Fandriks, L; Elfvin, A
BMC Gastroenterology null
Claire Anderson et al.
Nature communications, 7, 12656-12656 (2016-08-26)
Organizers are regions of the embryo that can both induce new fates and impart pattern on other regions. So far, surprisingly few organizers have been discovered, considering the number of patterned tissue types generated during development. This may be because
Prabha K Bista et al.
Microorganisms, 11(12) (2023-12-23)
Fusobacterium necrophorum, an anaerobic Gram-negative pathogen, causes necrotic cattle infections, impacting livestock health and the US feedlot industry. Antibiotic administration is the mainstay for treating F. necrophorum infections, although resistance hampers their effectiveness. Vaccination, especially targeting outer membrane proteins (OMPs)
Fang Ding et al.
PloS one, 10(5), e0123939-e0123939 (2015-05-07)
'Candidatus Liberibacter asiaticus' (CaLas), a non-cultured member of the α-proteobacteria, is the causal agent of citrus Huanglongbing (HLB). Due to the difficulties of in vitro culture, antibodies against CaLas have not been widely used in studies of this pathogen. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service