Skip to Content
Merck
All Photos(1)

Documents

1.07229

Millipore

Proteose peptone

suitable for microbiology

Synonym(s):

Proteose peptone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106212
EC Index Number:
293-426-3
NACRES:
NA.74

biological source

animal

Quality Level

form

powder

pH

6.5-7.5 (20 g/L in H2O)

solubility

500 g/L

bulk density

250 kg/m3

application(s)

microbiology

storage temp.

15-25°C

Analysis Note

pH-value (2 %; water): 6.5 - 7.5
Heavy metals (as Pb): ≤ 0.001 %
Sulfated ash (600 °C): ≤ 15 %
Loss on Drying (105°C): ≤ 10 %
Nitrogen (calculated on dried substance): ≥ 12.0 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service