Skip to Content
Merck
All Photos(3)

Key Documents

P5905

Sigma-Aldrich

Peptone from animal tissue

from meat, BioReagent, suitable for cell culture, suitable for plant cell culture

Synonym(s):

Peptone from animal source

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42040102
NACRES:
NA.75

biological source

animal (meat)

product line

BioReagent

form

powder

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

solubility

H2O: 50 mg/mL

shipped in

ambient

storage temp.

room temp

SMILES string

O(C)C(=O)C(CCC(CC)(CC)C)C(=O)OC

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Peptone is a vital ingredient of the culture medium, which is produced by the digestion of lean meat with proteolytic enzymes. It is a complex mixture of partially metabolised proteins. Peptone is a source of nutrition for bacterial growth.

Application

Peptone from animal tissue has been used as a reagent in culture medium. It has also been used as a stimulus for luminal perfusion.
To study its effect on glucagon-like peptide-1 (GLP-1) secretion from murine primary colonic cultures and Ca2+ dynamics in L cells.

Other Notes

Enzymatic hydrolysate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Textbook of Microbiology & Immunology - E-book (2014)
Eleftheria Diakogiannaki et al.
Diabetologia, 56(12), 2688-2696 (2013-09-21)
Ingested protein is a well-recognised stimulus for glucagon-like peptide-1 (GLP-1) release from intestinal L cells. This study aimed to characterise the molecular mechanisms employed by L cells to detect oligopeptides. GLP-1 secretion from murine primary colonic cultures and Ca(2+) dynamics
Yeast transformation efficiency is enhanced by TORC 1-and eisosome-dependent signaling
Yu SC, et al.
MicrobiologyOpen, e00730-e00730 (2018)
Intracellular sorting and transcytosis of the rat transferrin receptor antibody OX26 across the blood-brain barrier in vitro is dependent on its binding affinity
Haqqani AS, et al.
Journal of Neurochemistry, 146(6), 735-752 (2018)
Establishment of a Symbiotic in vitro System between a Green Meadow Orchid and a Rhizoctonia-like Fungus
Ercole E, et al.
Bio-protocol, 5(3), 1482-1482 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service