Skip to Content
Merck
All Photos(3)

Key Documents

B4888

Millipore

Beef Extract for Biotechnology and Microbiology

for Biotechnology and Microbiology

Synonym(s):

Meat extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.74
Pricing and availability is not currently available.

Product Name

Beef extract powder, suitable for microbiology, dehydrated extract of bovine tissue used in the preparation of microbiological culture media

biological source

bovine skeletal muscle

Quality Level

sterility

non-sterile

form

powder

nitrogen analysis

~12% total

impurities

<9% sodium chloride

loss

≤7% loss on drying

color

yellow-brown

pH

6.5-7.5

solubility

H2O: soluble

application(s)

food and beverages
microbiology

General description

Beef extract powder is a dehydrated extract of bovine tissue for use in preparing microbiological culture media. It is prepared from selected bovine tissues which are free from fat and sinew. Diverse media utilize beef extract, for example, yeast dextrose broth, Herrold′s egg yolk medium. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.

Application

Beef Extract Powder is used in general purpose culture media and is recommended for use in the bacteriological examination of water, milk, and other materials, where the uniform composition of media is important.Beef Extract Powder is also relied upon for biochemical studies like fermentation reactions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation of sabin-like polioviruses from wastewater in a country using inactivated polio vaccine
Zurbriggen S, et al.
Applied and Environmental Microbiology, 74(18), 5608- 5614 (2008)
Sebastian Zurbriggen et al.
Applied and environmental microbiology, 74(18), 5608-5614 (2008-07-22)
From 2001 to 2004, Switzerland switched from routine vaccination with oral polio vaccine (OPV) to inactivated polio vaccine (IPV), using both vaccines in the intervening period. Since IPV is less effective at inducing mucosal immunity than OPV, this change might
Highly efficient Agrobacterium-based transformation system for callus cells of the C 3 halophyte Suaeda salsa
Zhao SZ, et al.
Acta Physiologiae Plantarum / Polish Academy of Sciences, Committee of Plant Physiology Genetics and Breeding, 30(5), 729- 736 (2008)
Preparation, application and testing of permanent antibacterial and antiviral coatings
Haldar J, et al
Nature Protocols, 2(10), 2412-2412 (2007)
Reduction of Mycobacterium avium ssp. paratuberculosis in colostrum: Development and validation of 2 methods, one based on curdling and one based on centrifugation
Verhegghe M, et al.
Journal of Dairy Science, 100(5), 3497- 3512 (2017)

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service