1.06727
Nickel(II) sulfate hexahydrate
for analysis EMSURE® ACS
Synonym(s):
Nickel(II) sulfate hexahydrate
About This Item
Recommended Products
grade
ACS reagent
Quality Level
product line
EMSURE®
Assay
99.0-101.0% (complexometric)
form
crystals
potency
361 mg/kg LD50, oral (Rat)
impurities
≤0.005% Insoluble matter
pH
4.3-4.7 (20 °C, 100 g/L in H2O)
solubility
650 g/L
density
2.07 g/cm3 at 20 °C
bulk density
1000 kg/m3
anion traces
chloride (Cl-): ≤0.001%
cation traces
Ca: ≤0.005%
Co: ≤0.002%
Cu: ≤0.002%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
Mn: ≤0.002%
N: ≤0.001%
Na: ≤0.03%
Pb: ≤0.001%
Zn: ≤0.002%
storage temp.
2-30°C
SMILES string
[Ni+2].[S](=O)(=O)([O-])[O-].O.O.O.O.O.O
InChI
1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2
InChI key
RRIWRJBSCGCBID-UHFFFAOYSA-L
Related Categories
Analysis Note
Identity: passes test
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.001 %
Total nitrogen (N): ≤ 0.001 %
Ca (Calcium): ≤ 0.005 %
Co (Cobalt): ≤ 0.002 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.005 %
Mn (Manganese): ≤ 0.002 %
Na (Sodium): ≤ 0.03 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.002 %
Legal Information
Signal Word
Danger
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation
Target Organs
Respiratory Tract
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service