1.05941
Manganese(II) sulfate monohydrate
spray-dried for analysis EMSURE® ACS,Reag. Ph Eur
Synonym(s):
Manganese(II) sulfate monohydrate
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
product line
EMSURE®
Assay
98.0-101.0% (MnSO4.H2O basis, complexometric)
99.0-101.0% (calc. on the ignited basis, complexometric)
form
solid
potency
2150 mg/kg LD50, oral (Rat)
impurities
≤0.01% Insoluble matter
loss
10.0-12.0% loss on ignition, 500 °C
pH
3.0-3.5 (20 °C, 50 g/L in H2O)
mp
>449 °C (anhydrous substance)
solubility
762 g/L
density
2.95 g/cm3 at 20 °C
bulk density
1000‑1200 kg/m3
anion traces
chloride (Cl-): ≤0.001%
cation traces
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.001%
Zn: ≤0.005%
heavy metals (as Pb): ≤0.001%
storage temp.
15-25°C
SMILES string
[Mn+2].[S](=O)(=O)([O-])[O-].O
InChI
1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2
InChI key
ISPYRSDWRDQNSW-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Linkage
Analysis Note
Assay (complexometric, calc. on the ignited basis): 99.0 - 101.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.005 %
Ni (Nickel): ≤ 0.0005 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.005 %
Substances reducing potassium permanganate: passes test
Loss on ignition (500 °C): 10.0 - 12.0 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation
Target Organs
Brain
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service