Skip to Content
Merck
All Photos(1)

Key Documents

31462

Sigma-Aldrich

Nickel(II) chloride hexahydrate

puriss. p.a., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2 · 6H2O
CAS Number:
Molecular Weight:
237.69
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
form:
powder or crystals
Pricing and availability is not currently available.

grade

puriss. p.a.

Quality Level

Assay

≥98%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: nickel

pH

4-7 (20 °C, 5%)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤10 mg/kg
Co: ≤100 mg/kg
Cu: ≤10 mg/kg
Fe: ≤20 mg/kg
Mg: ≤10 mg/kg
Na: ≤50 mg/kg
Pb: ≤10 mg/kg
Zn: ≤10 mg/kg

storage temp.

room temp

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ni]Cl

InChI

1S/2ClH.Ni.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

LAIZPRYFQUWUBN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) chloride hexahydrate is a hydrated nickel salt. Its crystals were monoclinic and structure was investigated by X-ray single crystal method.[1]

Application

Nickel(II) chloride hexahydrate may be employed as a catalyst in the high yield synthesis of tetra-substituted pyrrole derivatives.[2] It may be used for the conversion of tetrahydropyranyl (THP) and tert-butyldimethylsilyl (TBS) ethers to the corresponding hydroxyl compounds.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Crystal Structure of Nickel Chloride Hexahydrate, NiCl2.6H2O.
Mizuno J.
Journal of the Physical Society of Japan, 16(8), 1571-1580 (1961)
Synthesis of tetra-substituted pyrroles, a potential phosphodiesterase 4B inhibitor, through nickel (II) chloride hexahydrate catalyzed one-pot four-component reaction.
Khan AT, et al.
Tetrahedron Letters, 53(32), 4145-4150 (2012)
A catalytic amount of nickel (II) chloride hexahydrate and 1, 2-ethanedithiol is a good combination for the cleavage of tetrahydropyranyl (THP) and tert-butyldimethylsilyl (TBS) ethers.
Khan AT, et al.
Tetrahedron Letters, 45(52), 9617-9621 (2004)
Rebekka Kubisch et al.
Sensors (Basel, Switzerland), 12(3), 3370-3393 (2012-06-28)
Pollution of drinking water sources represents a continuously emerging problem in global environmental protection. Novel techniques for real-time monitoring of water quality, capable of the detection of unanticipated toxic and bioactive substances, are urgently needed. In this study, the applicability
Ritu Tyagi et al.
Journal of applied toxicology : JAT, 33(2), 134-141 (2011-09-20)
Heavy metals are known for their associated nephrotoxicity and nickel is no exception. An integrated metabonomic approach, based on high-resolution (1) H NMR spectroscopy, was applied to determine the acute biochemical effects of NiCl(2) on the renal tissues of rats.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service