Skip to Content
Merck
All Photos(6)

Key Documents

06-599

Sigma-Aldrich

Anti-acetyl-Histone H3 Antibody

from rabbit

Synonym(s):

H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

ChIP: suitable (ChIP-seq)
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (not specified)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)
rat ... H3C1(679994)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Acetylation of histone H3 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs). Acetylation of lysine14 is commonly seen in genes that are being actively transcribed into RNA.

Specificity

Broad species cross-reactivity expected based on sequence similarity.
Recognizes Histone H3 acetylated at the N-terminus.

Immunogen

Epitope: N-terminus.
Linear peptide corresponding to human Histone H3 acetylated at the N-terminus.

Application

Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
Immunocytochemistry Analysis: 1:500 dilution of a representative lot of this antibody detected acetylated Histone H3 in NIH/3T3 and A431 cells.

Chromatin Immunoprecipitation: This antibody has been shown to work for chIP using HeLa extracted chromatin and 5 mg of this antibody.

ChIP-Seq: This antibody has been shown to work for ChIP-Seq in Kanai, A., et. al. (2011). DNA Research 18:379-392.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blot in HeLa acid extract, sodium butyrate treated HeLa cells, and recombinant Histone H3.

Western Blot Analysis: 0.05 µg/mL of this antibody detected Histone H3 on 10 µg of HeLa acid extract, sodium butyrate treated HeLa cells, and recombinant Histone H3.

Target description

~ 17 kDa observed

Physical form

Format: Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract, sodium butyrate treated HeLa cells, and recombinant Histone H3

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differences in stability of repressor complexes at promoters underlie distinct roles for Rb family members.
Young, AP; Longmore, GD
Oncogene null
Deletion of the major GATA1 enhancer HS 1 does not affect eosinophil GATA1 expression and eosinophil differentiation.
Guyot, B; Valverde-Garduno, V; Porcher, C; Vyas, P
Blood null
Differences in the chromatin structure and cis-element organization of the human and mouse GATA1 loci: implications for cis-element identification.
Valverde-Garduno, V; Guyot, B; Anguita, E; Hamlett, I; Porcher, C; Vyas, P
Blood null
Coregulator recruitment and histone modifications in transcriptional regulation by the androgen receptor.
Kang, Z; Janne, OA; Palvimo, JJ
Molecular Endocrinology null
Promoter-specific p53-dependent histone acetylation following DNA damage.
Kaeser, MD; Iggo, RD
Oncogene null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service