Skip to Content
Merck
All Photos(7)

Key Documents

07-449

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys27) Antibody

Upstate®, from rabbit

Synonym(s):

H3 histone family, member T, histone 3, H3, histone cluster 3, H3, H3k27me3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IHC
IP
WB
multiplexing
species reactivity:
human, mouse
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
multiplexing: suitable
western blot: suitable
citations:
2211
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

trimethylation (Lys27)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species cross-reactivity expected.
Trimethylated histone H3 (Lys27). Broad species cross-reactivity expected.

Immunogen

KLH-conjugated, synthetic 2X-branched peptide containing the sequence …AR(me3K)SAP… in which me3K corresponds to trimethyl-lysine at residue 27 of human histone H3.

Application

Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in WB, ICC, IP.
Immunocytochemistry:

Immunoprecipitation:

Beadlyte Histone-Peptide Specificity Assay:

Quality

Routinely evaluated by western blot in acid extracted proteins from HeLa cells.

Western Blot Analysis:
0.4 µg/mL of anti-trimethyl histone H3 Lys27 was used on recombinant Histone H3 and HeLa cell acid extracts.

Target description

17 kDa

Physical form

Format: Purified
Purified rabbit IgG in buffer containing 0.1 M Tris-Glycine, pH 7.4, 0.15 M NaCl with 0.05% sodium azide before the addition of 30% glycerol.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An extended transcriptional network for pluripotency of embryonic stem cells.
Kim, J; Chu, J; Shen, X; Wang, J; Orkin, SH
Cell null
A WD40 domain cyclophilin interacts with histone H3 and functions in gene repression and organogenesis in Arabidopsis.
Li, H; He, Z; Lu, G; Lee, SC; Alonso, J; Ecker, JR; Luan, S
Plant Cell null
Su(z)2 antagonizes auto-repression of Myc in Drosophila, increasing Myc levels and subsequent trans-activation.
Khan, A; Shover, W; Goodliffe, JM
Testing null
Polycomb-group proteins repress the floral activator AGL19 in the FLC-independent vernalization pathway.
Schonrock, N; Bouveret, R; Leroy, O; Borghi, L; Kohler, C; Gruissem, W; Hennig, L
Genes & Development null
COOH-terminal binding protein regulates expression of the p16INK4A tumor suppressor and senescence in primary human cells.
Mroz, EA; Baird, AH; Michaud, WA; Rocco, JW
Cancer Research null

Articles

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

See All

Protocols

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service