Skip to Content
Merck
All Photos(2)

Key Documents

830845P

Avanti

14:0 PA

1,2-dimyristoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1,2-ditetradecanoyl-sn-glycero-3-phosphate (sodium salt); DMPA; PA(14:0/14:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H60O8PNa
CAS Number:
Molecular Weight:
614.77
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (830845P-200mg)
pkg of 1 × 25 mg (830845P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 830845P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCC)O

InChI

1S/C31H61O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-30(32)37-27-29(28-38-40(34,35)36)39-31(33)26-24-22-20-18-16-14-12-10-8-6-4-2;/h29H,3-28H2,1-2H3,(H2,34,35,36);/q;+1/p-1/t29-;/m1./s1

InChI key

UBSPGYHFNIKQIP-XXIQNXCHSA-M

General description

Phosphatidic acid is obtained from glycerophospholipid with the help of phospholipase D. 14:0 PA/a negatively charged 1,2-dimyristoyl-sn-glycero-3-phosphate (DMPA) carries a phosphoric acid head group.

Application

14:0 PA may be used to prepare liposomes. It may also be used as a phospholipid (PL) standard to quantify phosphatidic acid (PA).

Biochem/physiol Actions

Phosphatidic acid might serve as a modulator of phagocyte respiratory burst. It also acts as a precursor of diacylglycerol and lysophosphatidic acid.

Packaging

5 mL Amber Glass Screw Cap Vial (830845P-200mg)
5 mL Amber Glass Screw Cap Vial (830845P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjie Wang et al.
The journal of physical chemistry. B, 116(24), 7213-7220 (2012-05-23)
Although a minor component of the lipidome, phosphatidic acid (PA) plays a crucial role in nearly all signaling pathways involving cell membranes, in part because of its variable electrical charge in response to environmental conditions. To investigate how charge is
Extraction and profiling of plant polar glycerol lipids
Liu, Yu and Wang, Xuemin
Bio-protocol (2016)
Encapsulation and release of a hydrophobic drug from hydroxyapatite coated liposomes
Xu Q, et al.
Biomaterials, 28(16), 2687-2694 (2007)
Chiaki Murakami et al.
The Journal of biological chemistry, 295(10), 2932-2947 (2020-01-26)
The δ isozyme of diacylglycerol kinase (DGKδ) plays critical roles in lipid signaling by converting diacylglycerol (DG) to phosphatidic acid (PA). We previously demonstrated that DGKδ preferably phosphorylates palmitic acid (16:0)- and/or palmitoleic acid (16:1)-containing DG molecular species, but not
Glycerolipids in signal transduction
Lambeth JD and Ryu SH
New comprehensive biochemistry, 31, 237-255 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service