Skip to Content
Merck
All Photos(2)

Key Documents

S7400

Sigma-Aldrich

Sodium (meta)arsenite

≥90%

Synonym(s):

Arsenenous acid sodium salt, Arsenic sodium oxide, Arsenite sodium, Sodium arsenic oxide, sodium arsenite, Sodium dioxoarsenate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaAsO2
CAS Number:
Molecular Weight:
129.91
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥90%

reaction suitability

core: arsenic
reagent type: catalyst

solubility

H2O: 10%, clear to turbid

SMILES string

[Na+].[O-][As]=O

InChI

1S/AsHO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

PTLRDCMBXHILCL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium (meta) arsenite can be used as a precursor in the synthesis of nano sized arsenic imprinted polymer which is used as a sensor in the determination of arsenic(As3+).[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruiqi Wang et al.
Cell death & disease, 10(3), 215-215 (2019-03-06)
Stress granule formation is important for stress response in normal cells and could lead to chemotherapy resistance in cancer cells. Aberrant stress granule dynamics are also known to disrupt proteostasis, affect RNA metabolism, and contribute to neuronal cell death. Meanwhile
Gergely Jocsak et al.
Brain sciences, 9(12) (2019-12-11)
The endocrine system of animals consists of fine-tuned self-regulating mechanisms that maintain the hormonal and neuronal milieu during tissue development. This complex system can be influenced by endocrine disruptors (ED)-substances that can alter the hormonal regulation even in small concentrations.
Martin P Pothier et al.
Frontiers in microbiology, 9, 2310-2310 (2018-10-20)
Despite its high toxicity and widespread occurrence in many parts of the world, arsenic (As) concentrations in decentralized water supplies such as domestic wells remain often unquantified. One limitation to effective monitoring is the high cost and lack of portability
Andrea Maria Guarino et al.
Genes, 9(10) (2018-10-27)
The prototype cold-shock Y-box binding protein 1 (YB-1) is a multifunctional protein that regulates a variety of fundamental biological processes including cell proliferation and migration, DNA damage, matrix protein synthesis and chemotaxis. The plethora of functions assigned to YB-1 is
Angel Guerra-Moreno et al.
FEBS letters, 593(10), 1080-1088 (2019-04-20)
In the unfolded protein response (UPR), Ire1 activates Hac1 to coordinate the transcription of hundreds of genes to mitigate ER stress. Recent work in Caenorhabditis elegans suggests that oxidative stress inhibits this canonical Ire1 signalling pathway, activating instead an antioxidant

Protocols

Enzymatic Assay of Neuraminidase applies to products that have a specification for neuraminidase content by enzymatic determination.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service