Skip to Content
Merck
All Photos(4)

Key Documents

S2002

Sigma-Aldrich

Sodium azide

ReagentPlus®, ≥99.5%

Synonym(s):

Hydrazoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaN3
CAS Number:
Molecular Weight:
65.01
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (inorganic)

Quality Level

product line

ReagentPlus®

Assay

≥99.5%

form

powder

storage condition

(Tightly closed. Dry. Keep in a well-ventilated place. Do not store near acids.)

technique(s)

cell based assay: suitable
immunohistochemistry: suitable

color

white

mp

370-425 °C

solubility

soluble 65 g/L at 20 °C (completely)

suitability

suitable for chromatography
suitable for derivatization

application(s)

sample preparation

SMILES string

[Na]N=[N+]=[N-]

InChI

1S/N3.Na/c1-3-2;/q-1;+1

InChI key

PXIPVTKHYLBLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium Azide (NaN3) is a potent inhibitor – a water-soluble crystalline powder that′s odorless. Widely recognized for its versatile applications, it plays a pivotal role across molecular biology, cell culture, and biochemical research. Sodium azide is commonly used as a bacteriostatic preservative in aqueous laboratory reagents and biological fluids. It is also a metabolic inhibitor, which inhibits oxidative phosphorylation. In cell culture, it maintains the integrity of your cell lines by preventing microbial contamination. Its application extends to molecular biology, where it aids in preserving nucleic acids for downstream analysis, ensuring the fidelity of your results.

Application

Baylis-Hillman acetates such as ethyl 2-(acetoxyphenylmethyl)acrylate can undergo nucleophilic displacement by sodium azide in aqueous medium to form ethyl (E)-2-azidomethyl-3-phenylpropenoate.
Sodium azide is suitable for use in histopathology to prepare and store tissue samples. Also used as a component of staining buffer (HEPES buffer) for whole mount immunolabelling.
Catalyst for:

  • Oxidative decarboxylation
  • Michael addition reactions

Reagent for synthesis of
  • Blue fluorescent copolymers
  • Metal phosphonates
  • Arenes via aminations

Biochem/physiol Actions

Sodium azide is clinically used as a preservative in diluting fluid for counting red blood cells. It prevents capping and internalization of fluorescent surface-bound antibodies in various scientific applications, including flow cytometry.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

comparable product

Product No.
Description
Pricing

suggested gloves for splash protection

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium uptake by rat kidney mitochondria.
DeLuca HF and Engstrom GW.
Proceedings of the National Academy of Sciences of the USA, 47(11), 1744-1750 (1961)
A critical review of published methods for analysis of red cell antigen-antibody reactions by flow cytometry, and approaches for resolving problems with red cell agglutination
Patricia A Arndt , George Garratty
Transfusion Medicine Reviews, 24(3), 174-192 (2010)
Sodium azide, a bacteriostatic preservative contained in commercially available laboratory reagents, influences the responses of human platelets via the cGMP/PKG/VASP pathway.
Russo I, et al.
Clinical Biochemistry, 41(4), 343-349 (2008)
Nucleophilic displacement by azide and cyanide on Baylis-Hillman acetates in water.
Yadav JS, et al.
Tetrahedron Letters, 46(16), 2761-2763 (2005)
Abris Gilvesy et al.
Acta neuropathologica, 144(4), 651-676 (2022-08-31)
Tau pathology of the noradrenergic locus coeruleus (LC) is a hallmark of several age-related neurodegenerative disorders, including Alzheimer's disease. However, a comprehensive neuropathological examination of the LC is difficult due to its small size and rod-like shape. To investigate the

Protocols

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service