Skip to Content
Merck
All Photos(1)

Key Documents

M4128

Sigma-Aldrich

Mercury(II) thiocyanate

96.5-103.5% (titration)

Synonym(s):

Mercuric isothiocyanate, Mercuric sulfocyanide, Mercuric thiocyanate, Mercury di(thiocyanate), Thiocyanic acid, mercury(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(SCN)2
CAS Number:
Molecular Weight:
316.75
Beilstein:
3561344
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96.5-103.5% (titration)

reaction suitability

core: mercury
reagent type: catalyst

mp

165 °C (dec.) (lit.)

density

3.71 g/mL at 25 °C (lit.)

SMILES string

N#CS[Hg]SC#N

InChI

1S/2CHNS.Hg/c2*2-1-3;/h2*3H;/q;;+2/p-2

InChI key

GBZANUMDJPCQHY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Mercury(II) thiocyanate is the coordination complex of Hg2+ and the thiocyanate ion(SCN-). It is primary used as catalyst for the addition of thiocyanic acid (HSCN) across the triple bond of a variety of alkynes and as a precursor to synthesize other thiocyanate complexes. Within analytical chemistry, it is also used for the colorimetric determination of chloride ions in water.

Application

Mercury(II) thiocyanate can be employed as a reactant for the synthesis of:

  • Polymeric mercury(II) coordination complexes with various derivatives of 4,4′-bipyridine ligands via the branched tube method.
  • Manganese mercury thiocyanate (MMTC), an organometallic optical single-crystal.
  • Heterotrinuclear copper(II)–mercury(II)–copper(II)complexes possessing antibacterial and antifungal activities.

Quality

Contains a small amount of mercurous thiocyanate insoluble in dilute acid.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mercury(II) Thiocyanate.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Surface modification of chromatography adsorbents by low temperature low pressure plasma
A Arpanaei, et al.
Journal of Chromatography A, 6905-6916 (2010)
Photometric Determination of Chlorides in Water
David M. Zall, et al.
Analytical Chemistry, 1665-1668 (1956)
Mercury (II) acetate/thiocyanate coordination polymers with n-donor ligands, spectroscopic, thermal and structural studies.
Mahmoudi G , et al.
Inorgorganica Chimica Acta, 362(1), 217-225 (2009)
Solid-State 199Hg MAS NMR Studies of Mercury(II) Thiocyanate Complexes and Related Compounds. Crystal Structure of Hg(SeCN)2
Graham A. Bowmaker, et al.
Inorganic Chemistry, 37(8), 1734-1743 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service