Skip to Content
Merck
All Photos(6)

Key Documents

222488

Sigma-Aldrich

N-(1-Naphthyl)ethylenediamine dihydrochloride

ACS reagent, >98%

Synonym(s):

2-(1-Naphthylamino)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
259.17
Beilstein:
3707471
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

>98%

form

solid

impurities

≤5% water

mp

194-198 °C (dec.) (lit.)

solubility

H2O: passes test

suitability

passes test for sensitivity to sulfanilamide

functional group

amine

SMILES string

Cl.Cl.NCCNc1cccc2ccccc12

InChI

1S/C12H14N2.2ClH/c13-8-9-14-12-7-3-5-10-4-1-2-6-11(10)12;;/h1-7,14H,8-9,13H2;2*1H

InChI key

MZNYWPRCVDMOJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-(1-Naphthyl)ethylenediamine dihydrochloride (NEDA·2HCl) acts as donor molecule and forms charge transfer (CT) complexes with π-acceptors (quinol and picric acid), σ-acceptors (iodine) and vacant orbital acceptors (tin(IV) tetrachloride and zinc chloride). These complexes were investigated by positron annihilation lifetime spectroscopy (PAL).

Application

N-(1-Naphthyl)ethylenediamine dihydrochloride has been used for the preparation of staining solution for the visualization of the reaction product(s) of starch active enzymes, containing α-1, 4 linked and α-1, 6 linked glucose by thin layer chromatography. It has also been used in modified Saville and Griess assays.
N-(1-Naphthyl)ethylenediamine dihydrochloride may be used in the synthesis of 5-[1,2]dithiolan-3-yl-pentanoic acid [2-(naphthalene-1-ylamino)-ethyl]amide (DPAN) by reacting with (±)-1,2-dithiolane-3-pentanoic acid in the presence of N-(3-dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride and N-hydroxysuccinimide. The solution formed by dissolving N-(1-naphthyl)ethylenediamine dihydrochloride in sulfuric acid-methanol may be used as a reagent for the detection of sugars at nanoscale on thin-layer plates.
Coupling agent for the spectrophotometric determination of aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols, and sulfonamides.
Solution of N-(1-naphthyl)ethylenediamine dihydrochloride in sulfuric acid-methanol may be employed for the quantitation of sugars.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Product Analysis of Starch Active Enzymes by TLC.
Cockburn D and Koropatkin N.
Bio-protocol, 5(20) (2015)
Chi-Su Yoon et al.
Molecules (Basel, Switzerland), 23(9) (2018-09-20)
Nardostachys jatamansi contains various types of sesquiterpenoids that may play an important role in the potency of plant's anti-inflammatory effects, depending on their structure. In this study, five new sesquiterpenoids, namely kanshone L (1), kanshone M (2), 7-methoxydesoxo-narchinol (3), kanshone
K K Verma et al.
Talanta, 38(3), 283-289 (1991-03-01)
A general assay procedure for a wide variety of thiols is described. The technique has three steps: (1) formation of S-nitrosothiols with nitrous acid, (2) destruction of the excess of nitrous acid, (3) hydrolysis of the S-nitrosothiols with mercuric ions
Analytical Chemistry, 55, 1226-1226 (1983)
N-(1-naphthyl)ethylenediamine dihydrochloride as a new reagent for nanomole quantification of sugars on thin-layer plates by a mathematical calibration process.
M Bounias
Analytical biochemistry, 106(2), 291-295 (1980-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service