Skip to Content
Merck
All Photos(1)

Key Documents

E19154

Sigma-Aldrich

Ethylcyclohexane

≥99%

Synonym(s):

ECH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11C2H5
CAS Number:
Molecular Weight:
112.21
Beilstein:
1900337
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

25 mmHg ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

504 °F

expl. lim.

6.6 %

refractive index

n20/D 1.432 (lit.)

bp

130-132 °C (lit.)

mp

−111 °C (lit.)

density

0.788 g/mL at 25 °C (lit.)

SMILES string

CCC1CCCCC1

InChI

1S/C8H16/c1-2-8-6-4-3-5-7-8/h8H,2-7H2,1H3

InChI key

IIEWJVIFRVWJOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethylcyclohexane can be used as:
  • A model for alkyl-substituted cycloalkanes to study their rate coefficients of reaction with OH radicals and Cl atoms.
  • A model for light hydrocarbons to investigate their catalytic cracking performance.
  • An organic monomer precursor to synthesize organic-inorganic hybrid polymer thin films by plasma enhanced chemical vapor deposition.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas-phase rate coefficients for a series of alkyl cyclohexanes with OH radicals and Cl atoms.
Bejan IG, et al.
International Journal of Chemical Kinetics (2018)
A Fuangswasdi et al.
Water research, 41(6), 1343-1349 (2007-02-03)
Mixtures of anionic and cationic surfactants exhibit synergistic behavior as evidenced by low critical micelle concentrations (CMC) of the mixed system, increased surface activity, and improved detergency performance. The adsorption of a single-head anionic surfactant, sodium dodecyl sulfate (SDS), in
The characteristics of organic-inorganic hybrid low-k thin films by PECVD.
Cho SJ, et al.
Surface and Coatings Technology, 202(22-23), 5654-5658 (2008)
Zhen Chen et al.
The Journal of chemical physics, 135(12), 124515-124515 (2011-10-07)
The dielectric relaxation behavior of ethylbenzene (EBZ) in its viscous regime is measured, and the glass transition temperature (T(g) = 116 K) as well as fragility (m = 98) are determined. While the T(g) of EBZ from this work is
Catalytic cracking of alkylcyclohexanes: Modeling the reaction pathways and mechanisms.
Watson BA, et al.
International Journal of Chemical Kinetics, 29(7), 545-560 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service