Skip to Content
Merck
All Photos(4)

Key Documents

C87851

Sigma-Aldrich

Copper(II) acetylacetonate

97%

Synonym(s):

2,4-Pentanedione copper(II) derivative, Bis(2,4-pentanedionato)copper(II), Cu(acac)2, Cupric acetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(C5H7O2)2
CAS Number:
Molecular Weight:
261.76
Beilstein:
4157957
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

form

powder

reaction suitability

core: copper
reagent type: catalyst

mp

284-288 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[Cu]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Cu/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

QYJPSWYYEKYVEJ-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Dextromethorphan as a moderator of reward deficiency: Research explored Dextromethorphan′s role in moderating reward deficiency associated with serotonin transporter availability in MDMA-treated animals, indicating its potential application in neuropsychopharmacology and addiction studies (Chiu et al., 2024).

  • Pharmacotherapies targeting GABA-Glutamate neurotransmission: A review discussed the use of Dextromethorphan among pharmacotherapies targeting GABA-glutamate neurotransmission for treatment-resistant depression, highlighting its role as an NMDA receptor antagonist and sigma-1 receptor agonist (Vecera et al., 2023).

  • Experience-dependent psychological effects: A double-blind study compared the psychological effects of Dextromethorphan and Psilocybin in healthy volunteers, examining their experience-dependent and enduring impacts, relevant in clinical psychology and psychedelic research (Mathai et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ik-Soo Shin et al.
The Analyst, 136(10), 2151-2155 (2011-03-25)
Though recently Ir(III) complexes have attracted much interest in electrochemiluminescent (ECL) analysis due to their high emission in various wavelengths, there were a few studies reported on its analytical applications. In this study, we evaluate the ECL from (pq)(2)Ir(acac) (pq
Chih-Chia Huang et al.
Chemical communications (Cambridge, England), (23)(23), 3360-3362 (2009-06-09)
A general approach involving a solvothermal method was developed to synthesize a series of silicate nanoshells (<100 nm) where both Gd silicate and Gd silicate:Eu nanoshells were further demonstrated to exhibit dual-modality MRI and optical imaging functions.
Gui-Ge Hou et al.
Chemical communications (Cambridge, England), 47(38), 10731-10733 (2011-08-27)
Two NbO-type MOFs based on ditopic pyridyl substituted diketonate ligands were reported. One exhibits a reversible SC-SC water encapsulation, while the other shows an interesting guest-driven luminescent property based on M(III) acetylacetonate (M = Eu and Fe) guest species.
Zhan'ao Tan et al.
Physical chemistry chemical physics : PCCP, 14(42), 14589-14595 (2012-09-28)
A solution-processed vanadium oxide (s-VO(x)) anode buffer layer on an indium-tin-oxide (ITO) electrode was used instead of PEDOT:PSS for improving the stability and photovoltaic performance of the polymer solar cells (PSCs). The s-VO(x) layer was prepared by spin-coating a vanadyl
Nan Tian et al.
Dalton transactions (Cambridge, England : 2003), 39(37), 8613-8615 (2010-08-19)
The design, synthesis, photophysical and significantly improved electrooptical properties of a series of red emitting cyclometalated iridium(iii) complexes containing carbazolyl-acetylacetonate ligands are described.

Articles

Copper metal deposition processes are an essential tool for depositing interconnects used in microelectronic applications, giving group 11 (coinage metals: Copper, Silver, and Gold) an important place in atomic layer deposition (ALD) process development.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service