Skip to Content
Merck
All Photos(1)

Key Documents

96738

Sigma-Aldrich

(R)-(+)-5,5′-Dichloro-2,2′-bis(diphenylphosphino)-6,6′-dimethoxy-1,1′-biphenyl

≥95.0% (H-NMR)

Synonym(s):

(+)-(5,5′-Dichloro-6,6′-dimethoxy-1,1′-biphenyl)-2,2′-diyl-bis(diphenylphosphine), (+)-2,2′-Bis(diphenylphosphino)-5,5′-dichloro-6,6′-dimethoxy-1,1′-biphenyl, (R)-(+)-Cl-MeO-BIPHEP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H30Cl2O2P2
CAS Number:
Molecular Weight:
651.50
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (H-NMR)

form

crystals

optical activity

[α]/D +59±4°, c = 1% in chloroform

InChI

1S/C38H30Cl2O2P2/c1-41-37-31(39)23-25-33(43(27-15-7-3-8-16-27)28-17-9-4-10-18-28)35(37)36-34(26-24-32(40)38(36)42-2)44(29-19-11-5-12-20-29)30-21-13-6-14-22-30/h3-26H,1-2H3

InChI key

GCBRCSSVOVNEIX-UHFFFAOYSA-N

Other Notes

Transition metal complexes are used as catalysts for asymmetric hydrogenation; Eur Pat Appl 749973, 1995 (20.06.1995); Eur Pat Appl 1160237 (18.05.2001).

Legal Information

produced by Bayer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Laue et al.
Patent null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service