Skip to Content
Merck
All Photos(1)

Key Documents

693014

Sigma-Aldrich

(S)-H8-BINAP

technical grade

Synonym(s):

(S)-(−)-2,2′-Bis(diphenylphospino)-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-binaphthyl, [(1S)-5,5′,6,6′,7,7′,8,8′-octahydro-[1,1′-binaphthalene]-2,2′-diyl]bis[diphenylphosphine]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H40P2
CAS Number:
Molecular Weight:
630.74
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

form

powder

SMILES string

C1CCc2c(C1)ccc(P(c3ccccc3)c4ccccc4)c2-c5c6CCCCc6ccc5P(c7ccccc7)c8ccccc8

InChI

1S/C44H40P2/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-12,19-26,29-32H,13-18,27-28H2

InChI key

ANSOKCGDSQQISA-UHFFFAOYSA-N

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

We present an article concerning BINAP/SEGPHOS® Ligands and Complexes.

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service