Skip to Content
Merck
All Photos(1)

Key Documents

682144

Sigma-Aldrich

(R)-(–)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane

96%

Synonym(s):

(R)-Phanephos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H34P2
CAS Number:
Molecular Weight:
576.65
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

optical activity

[α]/D -34±4°, c = 1 in chloroform

mp

222-226 °C

functional group

phosphine

SMILES string

P(c7ccccc7)(c6ccccc6)c1c2ccc(c1)CCc3c(cc(cc3)CC2)P(c5ccccc5)c4ccccc4

InChI

1S/C40H34P2/c1-5-13-35(14-6-1)41(36-15-7-2-8-16-36)39-29-31-21-25-33(39)27-23-32-22-26-34(28-24-31)40(30-32)42(37-17-9-3-10-18-37)38-19-11-4-12-20-38/h1-22,25-26,29-30H,23-24,27-28H2

InChI key

GYZZZILPVUYAFJ-UHFFFAOYSA-N

Application

(R)-(-)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane may be used as a ligand in the following processes:
  • Enantioselective reductive cyclization of 1,6-enynes via asymmetric hydrogenation in the presence of a rhodium catalyst to form alkylidene-substituted heterocycles.
  • Asymmetric hydroboration of 3,3-disubstituted cyclopropenes to form 2,2-disubstituted cyclopropyl boronates.
  • Asymmetric ring-opening reactions of azabenzonorbornadienes in the presence of zinc(II) triflate and palladium(II) acetate to form aminodihydronaphthalenes.
Efficient ligand for asymmetric hydrogentation of dehydroamino acids, methyl esters and keytones.

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US5874629 and any patents arising therefrom apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Diversity of copper (I) complexes showing thermally activated delayed fluorescence: basic photophysical analysis
Czerwieniec, Rafal, and Hartmut Yersin
Inorganic Chemistry, 54.9, 4322-4327 (2015)
Synthesis, structural analysis, and chiral investigations of some atropisomers with ee-tetrahalogeno-1, 3-butadiene core
Piron, Flavia, et al.
The Journal of Organic Chemistry, 74.23, 9062-9070 (2009)
Enantioselective Reductive Cyclization of 1, 6-Enynes via Rhodium-Catalyzed Asymmetric Hydrogenation: C- C Bond Formation Precedes Hydrogen Activation.
Jang HY, et al.
Journal of the American Chemical Society, 127(17), 6174-6175 (2005)
Amide bond formation via C (sp 3)?H bond functionalization and CO insertion
Liu, Huizhen, et al.
Chemical Communications (Cambridge, England), 50.3, 341-343 (2014)
Catalytic enantioselective hydroboration of cyclopropenes.
Rubina M, et al.
Journal of the American Chemical Society, 125(24), 7198-7199 (2003)

Articles

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service