Skip to Content
Merck
All Photos(1)

Key Documents

930989

Sigma-Aldrich

Magnesium chloride hexahydrate

99.999% (trace metals basis)

Synonym(s):

Magnesium dichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

Quality Level

Assay

99.999% (trace metals basis)

form

crystalline

mp

117 °C

solubility

H2O: soluble (highly soluble)
acetone: slightly soluble
alcohols: slightly soluble

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride hexahydrate is a colorless to white, crystalline salt. The salt is soluble in water and slightly soluble in ethanol, acetone, and pyridine. The hydrate loses water when heated to higher temperatures, but thermal dehydration to the anhydrous salt is not straightforward. Chemically, magnesium chloride is a weak Lewis acid. One of the most common and simplest routes to producing magnesium chloride is extraction from naturally occurring brine, for example by solar evaporation of sea water, which can be purified by solution chemistries.

Application

One major application of high-purity magnesium chloride is in the synthesis of layered double hydroxides (LDH), which are high-surface area ionic clays. LDHs are studied for water filtration, among other applications, and usually consist of magnesium and aluminum ions because of the low toxicity of these metal cations. LDHs are synthesized from chloride salts using co-precipitation reactions, ion-exchange processes, and urea methods. Another application of high-purity magnesium chloride is as a reagent to dope or substitute magnesium ions into solid-state materials and nanoparticles. For example, Mg-doping in ZnO has shown to modulate its band structure and Mg-doping in lithium nickel cobalt manganese oxide helps suppress cation mixing and enables longer cycle lifetimes for its use as a cathode material.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New Approach for Removing Anionic Organic Dyes from Wastewater Based on Electrostatically Driven Assembly
Sansuk, S., Srijaranai, S., et al.
Environmental Science & Technology, 50, 6477?6484-6477?6484 (2016)
Preparation of Molybdenum Disulfide Coated Mg/Al Layered Double Hydroxide Composites for Efficient Removal of Chromium(VI)
Wang, J., Want, P., et al.
ACS sustainable chemistry & engineering, 5, 7165?7174-7165?7174 (2017)
Band gap engineering and enhanced photoluminescence of Mg doped ZnO nanoparticles synthesized by wet chemical route
Arshad, M., Ansari, M.M., et al.
Journal of Luminescence, 161, 275-280 (2015)
Effect of Mg doping on the structural and electrochemical performance of LiNi0.6Co0.2Mn0.2O2 cathode materials
Huang, Z., Wang, Z., et al.
Electrochimica Acta, 795-802, 182-182 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service