Skip to Content
Merck
All Photos(3)

Documents

449571

Sigma-Aldrich

Silver chloride

AnhydroBeads, −10 mesh, 99.998% trace metals basis

Synonym(s):

AgCl (Silver monochloride), Silver(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgCl
CAS Number:
Molecular Weight:
143.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 912 °C)

Quality Level

product line

AnhydroBeads

Assay

99.998% trace metals basis

impurities

≤25.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

455 °C (lit.)

SMILES string

Cl[Ag]

InChI

1S/Ag.ClH/h;1H/q+1;/p-1

InChI key

HKZLPVFGJNLROG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Silver chloride (AgCl) can be used for a variety of applications such as: the coating on the silver wire that can be used as a reference electrode in electrochemistry, can be deposited on conducting material for stable photocatalysis under visible light

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design rules for donors in bulk-heterojunction solar cells?Towards 10% energy-conversion efficiency
Scharber MC, et al.
Advanced Materials, 18(6), 789-794 (2006)
Ag@ AgCl: a highly efficient and stable photocatalyst active under visible light
Wang P, et al.
Angewandte Chemie (International ed. in English), 47(41), 7931-7933 (2008)
Fabrication and characterization of visible-light-driven plasmonic photocatalyst Ag/AgCl/TiO2 nanotube arrays
Yu J, et al.
The Journal of Physical Chemistry C, 113(37), 16394-16401 (2009)
The silver chloride photoanode in photoelectrochemical water splitting
Schurch D, et al.
The Journal of Physical Chemistry B, 106(49), 12764-12775 (2002)
Shaokun Zhang et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(2), 739-744 (2012-12-19)
Silver-containing antimicrobial agents are used in various medical products. However, their toxicity to mammalian cells has not been sufficiently evaluated. Numerous studies have unveiled evidence of significant antimicrobial properties associated with Ag ions. In cell culture media or human body

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service