Skip to Content
Merck
All Photos(1)

Documents

928399

Sigma-Aldrich

Pd-PEPPSI-IPent Catalyst ChemBeads

Synonym(s):

Dichloro[1,3-bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II), [1,3-Bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)dichloropalladium(II), [1,3-Bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56Cl3N3Pd
CAS Number:
Molecular Weight:
791.67
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.82

Assay

~5 wt.% loading of catalyst

Quality Level

form

solid

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

extent of labeling

~5 wt. % loading of catalyst

InChI

1S/C35H52N2.C5H4ClN.2ClH.Pd/c1-9-26(10-2)30-19-17-20-31(27(11-3)12-4)34(30)36-23-24-37(25-36)35-32(28(13-5)14-6)21-18-22-33(35)29(15-7)16-8;6-5-2-1-3-7-4-5;;;/h17-24,26-29H,9-16H2,1-8H3;1-4H;2*1H;/q;;;;+2/p-2

InChI key

BCXSKTXOKALLAZ-UHFFFAOYSA-L

Application

Catalyst has been used for Stille coupling, Negishi coupling and Suzuki coupling reactions.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (732117)

Legal Information

PEPPSI is a trademark of Total Synthesis Ltd.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service