Skip to Content
Merck
All Photos(1)

Key Documents

901841

Sigma-Aldrich

N-Hydroxysuccinimide ester-poly(ethylene glycol)-b-poly(ε-caprolactone)

PEG average Mn 5,000, PCL average Mn 5,000

Synonym(s):

NHS ester PEG-PCL diblock copolymer, NHS ester-PEG-PCL, PEG-b-PCL, PEG-PCL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H14NO5S[C2H4O]n[C6H10O2]mH
UNSPSC Code:
12352005
NACRES:
NA.23

form

powder or solid

mol wt

PCL average Mn 5,000
PEG average Mn 5,000

color

white to off-white

PDI

≤1.3 (by GPC)

storage temp.

−20°C

Application

N-Hydroxysuccinimide ester-poly(ethylene glycol)-b-poly(ε-caprolactone) is a functionalized, amphiphilic, diblock copolymer composed of a hydrophilic PEG block and a hydrophobic PCL block. These biodegradable, biocompatible polymers can self-assemble to form nanoparticles, such as micelles and polymersomes, in both aqueous and non-aqueous media. Due to these properties, these polymers are widely used in polymeric nanoparticle formulation to achieve controlled and targeted delivery of therapeutic agents (e.g. APIs, genetic material, peptides, vaccines, and antibiotics). Additionally, well-defined nanoparticles with tunable size and properties can be prepared by altering the molecular weight ratios between hydrophilic and hydrophobic blocks, as well as by controlling formulation parameters. The NHS ester functional group on the PEG chain enables rapid and facile surface functionalization, allowing for these materials to be used in applications such as targeted drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Folate-functionalized polymeric micelles for tumor targeted delivery of a potent multidrug-resistance modulator FG020326.
Yang, et al.
Journal of Biomedical Materials Research Part A, 86(1), 48-60 (2008)
Cem Varan et al.
Beilstein journal of nanotechnology, 8, 1446-1456 (2017-09-14)
Background: Brain tumors are the most common tumors among adolescents. Although some chemotherapeutics are known to be effective against brain tumors based on cell culture studies, the same effect is not observed in clinical trials. For this reason, the development

Articles

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service