Skip to Content
Merck
All Photos(1)

Key Documents

41214

Sigma-Aldrich

O-[(N-Succinimidyl)succinyl-aminoethyl]-O′-methylpolyethylene glycol

2000, chemical modification reagent amine reactive

Synonym(s):

Polyethylene glycol, O-Methyl-O′-[(N-succinimidyl)succinyl-aminoethyl]polyethylene glycol 2′000, O-Methyl-O′-[2-(succinylamino)-ethyl]polyethylene glycol 2′000 N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

O-[(N-Succinimidyl)succinyl-aminoethyl]-O′-methylpolyethylene glycol 2′000,

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service