Skip to Content
Merck
All Photos(1)

Documents

86370

Sigma-Aldrich

Tellurium dioxide

≥97.0%

Synonym(s):

Telluria, Tellurium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TeO2
CAS Number:
Molecular Weight:
159.60
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥97.0%

form

powder

mp

733 °C (lit.)

density

5.67 g/mL at 25 °C (lit.)

SMILES string

O=[Te]=O

InChI

1S/O2Te/c1-3-2

InChI key

LAJZODKXOMJMPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tellurium dioxide (TeO2) is a ceramic material that can be used as a semiconducting oxide. It has a wide band gap and high mobility as determined by density functional theory (DFT) calculations. In bulk quantity, it exists in two polymorphs which include tetragonal α-TeO2 and orthorhombic β-TeO2.

Application

TeO2 can be potentially used in medical imaging and industrial monitoring processes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Lact. - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrathin tellurium dioxide: emerging direct bandgap semiconductor with high-mobility transport anisotropy
Guo S, et al.
Nanoscale, 10(18), 8397-8403 (2018)
Gamma radiation-induced changes in the electrical and optical properties of tellurium dioxide thin films
Arshak K and Korostynska O
IEEE Sensors Journal, 3(6), 717-721 (2003)
Garima Tripathi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 486-489 (2008-03-14)
The energy transfer in Er3+: Sm3+ codoped binary TeO2-Li2O (TLO) glass has been studied using 532 nm laser radiation on the basis of fluorescence intensity and the lifetime measurements. It is observed that the trace of erbium ion can be
G Bilir et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 314-321 (2011-09-20)
TeO(2)-CdF(2)-WO(3) glasses with various compositions and Er(3+) concentrations were prepared by conventional melting method. Their optical properties were studied by measuring the absorption, luminescence spectra and the decay patterns at room temperature. From the optical absorption spectra the Judd-Ofelt parameters
Sankha Chattopadhyay et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 67(10), 1748-1750 (2009-05-05)
A simple and inexpensive method for the separation of medically useful no-carrier-added (nca) iodine radionuclides from bulk amounts of irradiated tellurium dioxide (TeO(2)) target was developed. The beta(-) emitting (131)I radionuclide, produced by the decay of (131)Te through the (nat)Te(n

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service