Skip to Content
Merck
All Photos(2)

Documents

401587

Sigma-Aldrich

Bismuth(III) acetate

≥99.99% trace metals basis

Synonym(s):

Bismuth triacetate, Acetic acid bismuth(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3Bi
CAS Number:
Molecular Weight:
386.11
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.99% trace metals basis

form

solid

reaction suitability

core: bismuth
reagent type: catalyst

impurities

≤5% Bi2O3

SMILES string

CC(=O)O[Bi](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Bi/c3*1-2(3)4;/h3*1H3,(H,3,4);/q;;;+3/p-3

InChI key

WKLWZEWIYUTZNJ-UHFFFAOYSA-K

Application

  • Bismuth(III) acetate is employed as a starting material in the synthesis of bismuth(III) sulfide (Bi2S3) which can be used in solution-processable bulk heterojunction solar cells.
  • It is used in the preparation of gold-bismuth sulfide (Au–Bi2S3) heteronanostructures and bismuth titanate nanorods as photocatalysts.
  • It is also used in the synthesis of triarylbismuth compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bismuth titanate nanorods and their visible light photocatalytic properties.
Pei LZ, et al.
J. Alloy Compounds, 622(26), 254-261 (2015)
M Makrinich et al.
Solid state nuclear magnetic resonance, 92, 19-24 (2018-05-12)
Dipolar recoupling under magic-angle spinning allows to measure accurate inter-nuclear distances provided that the two interacting spins can be efficiently and uniformly excited. Alexander (Lex) Vega has shown that adiabatic transfers of populations in quadrupolar spins during the application of
Photocatalytic Au-Bi2S3 Heteronanostructures.
Manna G, et al.
Angewandte Chemie (International Edition in English), 53(26), 6743-6746 (2014)
Hybrid solution-processed bulk heterojunction solar cells based on bismuth sulfide nanocrystals.
Martinez L, et al.
Physical Chemistry Chemical Physics, 15(15), 5482-5487 (2013)
Ying Xie et al.
Nanoscale, 10(3), 1279-1285 (2018-01-03)
Herein, we report a strategy to construct a semi-hollow plasmonic nanocavity and grow ultrathin sulfide nanosheets inside. The competition and cooperation of Au deposition with Ag etching based on flat Ag nanoplates are proposed. For the establishment of the semi-hollow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service