Skip to Content
Merck
All Photos(2)

Key Documents

243450

Sigma-Aldrich

Tellurium dioxide

≥99%

Synonym(s):

Tellurium(IV) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TeO2
CAS Number:
Molecular Weight:
159.60
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99%

form

powder

mp

733 °C (lit.)

density

5.67 g/mL at 25 °C (lit.)

SMILES string

O=[Te]=O

InChI

1S/O2Te/c1-3-2

InChI key

LAJZODKXOMJMPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tellurium dioxide, also known as tellurium(IV) oxide, is a white, crystalline solid with a melting point of 1,130 °C and a boiling point of 1,850 °C. Tellurium dioxide is an important material in a number of applications, including as a catalyst in the synthesis of plastics resins, and pharmaceuticals, and as a component in tellurium-based alloys. Tellurium dioxide is typically made at high temperatures either by decomposition of tellurium(IV) chloride or tellurium(IV) sulfate in the presence of oxygen.

Application

Tellurium dioxide is used to prepare tellurium metal, telluric acid, and many tellurium salts. It may also be used in the preparation of Ag2Te nanoparticles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Lact. - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingjun Wu et al.
Frontiers in chemistry, 8, 84-84 (2020-03-21)
A systematic electrochemical study was conducted to investigate the reduction of tellurium (Te) in alkaline solutions. The effect of various parameters, including tellurite ion concentration, applied potential, and pH was investigated by both linear sweep voltammograms (LSVs) and electrochemical quartz
Mao Shen et al.
Nanoscale research letters, 8(1), 253-253 (2013-05-31)
A facile one-step synthesis of CdTe quantum dots (QDs) in aqueous solution by atmospheric microwave reactor has been developed using 3-mercaptopropionic acid reduction of TeO2 directly. The obtained CdTe QDs were characterized by ultraviolet-visible spectroscopy, fluorescent spectroscopy, X-ray powder diffraction
Garima Tripathi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 486-489 (2008-03-14)
The energy transfer in Er3+: Sm3+ codoped binary TeO2-Li2O (TLO) glass has been studied using 532 nm laser radiation on the basis of fluorescence intensity and the lifetime measurements. It is observed that the trace of erbium ion can be
Edmond P F Lee et al.
The Journal of chemical physics, 121(7), 2962-2974 (2004-08-05)
Ab initio calculations have been carried out on low-lying singlet and triplet states of TeO2 at different levels of theory with basis sets of up to the augmented-polarized valence-quintuple-zeta quality. Equilibrium geometrical parameters, harmonic vibrational frequencies, and relative electronic energies
G Bilir et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 314-321 (2011-09-20)
TeO(2)-CdF(2)-WO(3) glasses with various compositions and Er(3+) concentrations were prepared by conventional melting method. Their optical properties were studied by measuring the absorption, luminescence spectra and the decay patterns at room temperature. From the optical absorption spectra the Judd-Ofelt parameters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service