Skip to Content
Merck
All Photos(2)

Key Documents

856177

Sigma-Aldrich

Methyl viologen dichloride hydrate

98%

Synonym(s):

1,1′-Dimethyl-4,4′-bipyridinium dichloride, Gramoxone, Paraquat dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14Cl2N2 · xH2O
CAS Number:
Molecular Weight:
257.16 (anhydrous basis)
Beilstein:
3752782
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

>300 °C (lit.)

SMILES string

[Cl-].[Cl-].[H]O[H].C[n+]1ccc(cc1)-c2cc[n+](C)cc2

InChI

1S/C12H14N2.2ClH.H2O/c1-13-7-3-11(4-8-13)12-5-9-14(2)10-6-12;;;/h3-10H,1-2H3;2*1H;1H2/q+2;;;/p-2

InChI key

RLGVBTFWTZJYIL-UHFFFAOYSA-L

General description

Methyl viologen dichloride hydrate is a well-known electron acceptor since it is having similar reduction potential value compared to the conduction band (−0.44 V) of TiO2.

Application

Methyl viologen dichloride hydrate can be used as an electron transfer agent, which provides insights into the dynamics of the excited state of quantum dot materials.

It can also be used as a mediator in the synthesis of ultrasmall bimetallic PdAg nanocatalysts.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 33, 6677-6677 (1993)
Claudia Montllor-Albalate et al.
Redox biology, 21, 101064-101064 (2018-12-24)
Cu/Zn Superoxide Dismutase (Sod1) is a highly conserved and abundant metalloenzyme that catalyzes the disproportionation of superoxide radicals into hydrogen peroxide and molecular oxygen. As a consequence, Sod1 serves dual roles in oxidative stress protection and redox signaling by both
The Journal of Physical Chemistry, 98, 6377-6377 (1994)
Youjia Yu et al.
Frontiers in physiology, 11, 65-65 (2020-03-03)
Paraquat (PQ) is a widely used herbicide which can cause high mortality to humans. However, relatively few studies focus on metabolic feature of PQ intoxication for investigating the underlying mechanisms. Here we performed non-targeted metabolomics profiling of serum samples from
David B Hicks et al.
Microbiology (Reading, England), 165(9), 1001-1012 (2019-07-17)
Alkaliphilic Bacillus pseudofirmus OF4, which has a broad pH growth range of 7.5 to above 10.5, is yellow-pigmented due to carotenoids. Carotenoids contribute to membrane rigidity and can alleviate cellular oxidative stress. This study was undertaken to gain insight into

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service