Skip to Content
Merck
All Photos(1)

Key Documents

271845

Sigma-Aldrich

Benzyl viologen dichloride

97%

Synonym(s):

1,1′-Dibenzyl-4,4′-bipyridinium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22Cl2N2
CAS Number:
Molecular Weight:
409.35
Beilstein:
3811585
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

mp

260 °C (dec.) (lit.)

SMILES string

[Cl-].[Cl-].C(c1ccccc1)[n+]2ccc(cc2)-c3cc[n+](Cc4ccccc4)cc3

InChI

1S/C24H22N2.2ClH/c1-3-7-21(8-4-1)19-25-15-11-23(12-16-25)24-13-17-26(18-14-24)20-22-9-5-2-6-10-22;;/h1-18H,19-20H2;2*1H/q+2;;/p-2

InChI key

NLOIIDFMYPFJKP-UHFFFAOYSA-L

General description

Benzyl viologen dichloride (BVD) is an electrochromic indicator, which contains two chlorine atoms and an organic constituent attached with two nitrogen cations. In a redox reaction, BVD can be reduced in three states with different colors.

Application

BVD can be reduced to its neutral state by using sodium borohydride, which forms an n-type dopant for organic semiconductor based applications. It may be introduced to form a path for the transport of electrons from the thylakoid membranes to the mediator, which can be used in the fabrication of photo-driven bioanode. It also acts as a dopant with 2D layered materials to form an n-type chemical dopant that can be used to enhance the electrical properties of the triboelectric devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Construction of photo-driven bioanodes using thylakoid membranes and multi-walled carbon nanotubes.
Takeuchi R, et al.
Bioelectrochemistry, 122(19), 158-163 (2018)
Benzyl viologen as an n-type dopant for organic semiconductors.
Huseynova G, et al.
Organic Electronics (2018)
Interactions of benzyl viologen with surface-bound single-and double-stranded DNA.
Pang D and Abruna H
Analytical Chemistry, 72(19), 4700-4706 (2000)
Triboelectric Series of 2D Layered Materials.
Seol M, et al.
Advanced Materials, 30(39), 1801210-1801210 (2018)
Vayu Maini Rekdal et al.
Science (New York, N.Y.), 364(6445) (2019-06-15)
The human gut microbiota metabolizes the Parkinson's disease medication Levodopa (l-dopa), potentially reducing drug availability and causing side effects. However, the organisms, genes, and enzymes responsible for this activity in patients and their susceptibility to inhibition by host-targeted drugs are

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service