Skip to Content
Merck
All Photos(1)

Key Documents

791628

Sigma-Aldrich

Trimethylamine-d9 N-Oxide

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3D9NO
CAS Number:
Molecular Weight:
84.17
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

technique(s)

bio NMR: suitable

mass shift

M+9

SMILES string

[O-][N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C3H9NO/c1-4(2,3)5/h1-3H3/i1D3,2D3,3D3

InChI key

UYPYRKYUKCHHIB-GQALSZNTSA-N

Related Categories

General description

Trimethylamine-d9 is isotopically enriched molecule.

Application

Trimethylamine-d9 N-oxide is used in the stability of hydrogen bonding between peptide, water with trimethylamine N-oxide.It is also used as an internal standard for the quantification of trimethylamine N-oxide by GC- or LC-MS.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filip Meersman et al.
Biophysical journal, 97(9), 2559-2566 (2009-11-04)
Trimethylamine N-oxide (TMAO) is a naturally occurring osmolyte that stabilizes proteins, induces folding, and counteracts the denaturing effects of urea, pressure, and ice. To establish the mechanism behind these effects, isotopic substitution neutron-scattering measurements were performed on aqueous solutions of
Tsung-Jen Lin et al.
Nutrition (Burbank, Los Angeles County, Calif.), 66, 29-37 (2019-06-16)
A plant-based diet has been associated with a reduced risk of cardiovascular (CV) diseases. This study aimed to determine the levels and correlations of CV-related biomarkers and the beneficial role of dietary habits. A total of 63 healthy vegetarians (n = 32)
Xiang Zhou et al.
ESC heart failure, 7(1), 188-193 (2020-01-22)
Accumulating evidence has demonstrated that intestinal microbiota-dependent trimethylamine N-oxide (TMAO) is involved in the pathogenesis of various cardiovascular diseases. The present study was designed to investigate the prognostic value of TMAO in patients with chronic heart failure (CHF) after myocardial
Zeneng Wang et al.
Analytical biochemistry, 455, 35-40 (2014-04-08)
Trimethylamine-N-oxide (TMAO) levels in blood predict future risk for major adverse cardiac events including myocardial infarction, stroke, and death. Thus, the rapid determination of circulating TMAO concentration is of clinical interest. Here we report a method to measure TMAO in
Makiko Shimizu et al.
The Journal of toxicological sciences, 43(6), 387-393 (2018-06-08)
Medicinal carnitine-derived and dietary-derived malodorous trimethylamine and its non-malodorous metabolite trimethylamine N-oxide were historically regarded as nontoxic. Clinical and toxicological interest has recently arisen because of their potential association with atherosclerosis. We previously reported a human physiologically based pharmacokinetic (PBPK)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service