Skip to Content
Merck
All Photos(1)

Key Documents

492051

Sigma-Aldrich

Choline chloride-(trimethyl-d9)

98 atom % D

Synonym(s):

Choline-tri(methyl-d3) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2N(CD3)3Cl
CAS Number:
Molecular Weight:
148.68
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

form

solid

mp

302-305 °C (dec.) (lit.)

mass shift

M+9

SMILES string

[Cl-].[2H]C([2H])([2H])[N+](CCO)(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i1D3,2D3,3D3;

InChI key

SGMZJAMFUVOLNK-KYRNGWDOSA-M

Related Categories

General description

Choline chloride-trimethyl-d9 is an isotopic analogue of Choline chloride.

Application

Choline chloride-trimethyl-d9 can be used in the determination of betaines by fast atom bombardment mass spectrometry.It is also used to study the nanostructure and hydration of deep eutectic solvent.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Rhodes et al.
Plant physiology, 84(3), 781-788 (1987-07-01)
A rapid, sensitive, and selective method for the determination of betaines is described and discussed. The method entails derivatizing the quaternary ammonium compounds to increase their sensitivity to detection by fast atom bombardment mass spectrometry. Sensitivity of detection increases markedly
Nelli Keksel et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 54, 66-76 (2019-01-23)
Chronic stress, an important factor in the development of depressive disorders, leads to an increased formation of cortisol, which causes a hyperactivity of the hypothalamic-pituitary-adrenal (HPA) axis. In addition, cortisol mediates an adaptive effect on plasma membrane fluidity which may
Conor C Horgan et al.
Journal of materials chemistry. B, 8(20), 4447-4459 (2020-05-07)
Extracellular vesicles (EVs) are biologically-derived nanovectors important for intercellular communication and trafficking. As such, EVs show great promise as disease biomarkers and therapeutic drug delivery vehicles. However, despite the rapidly growing interest in EVs, understanding of the biological mechanisms that
Nicolas M B Brancucci et al.
Cell, 171(7), 1532-1544 (2017-11-14)
Transmission represents a population bottleneck in the Plasmodium life cycle and a key intervention target of ongoing efforts to eradicate malaria. Sexual differentiation is essential for this process, as only sexual parasites, called gametocytes, are infective to the mosquito vector. Gametocyte production
William J Quinn et al.
Cell reports, 33(11), 108500-108500 (2020-12-17)
Immune cell function is influenced by metabolic conditions. Low-glucose, high-lactate environments, such as the placenta, gastrointestinal tract, and the tumor microenvironment, are immunosuppressive, especially for glycolysis-dependent effector T cells. We report that nicotinamide adenine dinucleotide (NAD+), which is reduced to NADH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service