Skip to Content
Merck
All Photos(1)

Key Documents

92262

Sigma-Aldrich

Trimethylamine solution

43.0-49.0% in H2O (T)

Synonym(s):

N,N,N-Trimethylamine, N,N-Dimethylmethanamine, TMA solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein:
956566
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

43.0-49.0% in H2O (T)

density

0.83-0.88 g/mL at 20 °C

functional group

amine

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylamine solution may be used for the fabrication of polysulfone membranes by introducing benzyl trimethylammonium cation groups. These membranes are useful in the development of anion exchange membrane fuel cells (AEMFCs).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Using microbial desalination cells to reduce water salinity prior to reverse osmosis
Mehanna M
Energy & Environmental Science, 3(8), 1114-1120 (2010)
Crosslinked, epoxy-based anion conductive membranes for alkaline membrane fuel cells
Zhou J
Journal of Membrane Science, 350(1-2), 286-292 (2010)
M Al-Waiz et al.
Clinical pharmacology and therapeutics, 42(5), 588-594 (1987-11-01)
Trimethylamine (TMA) and its N-oxide (TMAO) are normal components of human urine. They are present in the diet and also derived from the enterobacterial metabolism of precursors such as choline. Dietary TMA is almost entirely metabolized to and excreted as
S L Hillier
American journal of obstetrics and gynecology, 169(2 Pt 2), 455-459 (1993-08-01)
Bacterial vaginosis is characterized microbiologically by replacement of the Lactobacillus-predominant vaginal flora by Gardnerella vaginalis, Bacteroides species, Mobiluncus species, and genital mycoplasmas. A standardized, laboratory-based diagnostic test for bacterial vaginosis is desirable in those instances in which a microscope is
Qian Li et al.
Current biology : CB, 23(1), 11-20 (2012-11-28)
Rodents use olfactory cues for species-specific behaviors. For example, mice emit odors to attract mates of the same species, but not competitors of closely related species. This implies rapid evolution of olfactory signaling, although odors and chemosensory receptors involved are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service