767972
Cell Free Amino Acid Mixture-15N
5-100 mM in water, 98 atom % 15N
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352209
NACRES:
NA.12
Recommended Products
isotopic purity
98 atom % 15N
Quality Level
form
solid
concentration
5-100 mM in water
technique(s)
bio NMR: suitable
protein expression: suitable
shipped in
dry ice
storage temp.
−20°C
Application
This product suitable as source of Stable Isotope Labeled amino acids for Cell Free Protein Synthesis. For Mass Spec applications, consider product 909653, Stable Isotope Labeled Amino Acid Mixture for Mass Spec.
The product may appear turbid, due to the relatively high concentration of tyrosine. Once diluted for use, solution should be clear.
The product may appear turbid, due to the relatively high concentration of tyrosine. Once diluted for use, solution should be clear.
Packaging
1 mL aqueous solution of all 20 amino acids.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Analysis Note
Amino acid concentrations vary by lot. Please refer to the specific lot Certificate of Analysis.
Approximate concentrations (mM):
ASP 60
THR 35
SER 35
GLU 40
PRO 20
GLY 100
ALA 100
VAL 40
MET 10
ILE 30
LEU 45
TYR 10
PHE 16
HIS 5
LYS 15
ARG 10
GLN 20
ASN 20
TRP 20
CYS 20
Approximate concentrations (mM):
ASP 60
THR 35
SER 35
GLU 40
PRO 20
GLY 100
ALA 100
VAL 40
MET 10
ILE 30
LEU 45
TYR 10
PHE 16
HIS 5
LYS 15
ARG 10
GLN 20
ASN 20
TRP 20
CYS 20
required but not provided
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Filippo Caschera et al.
BioTechniques, 58(1), 40-43 (2015-01-22)
Here we present a procedure for preparing amino acid mixtures--having both the desired composition and a physiological pH--at high concentrations for cell-free expression systems. Up to 2.1 mg/mL of active protein was synthesized in batch mode reactions with an all
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service